Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136946740-136946995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8G
Entrez Gene ID
733
Gene Name
complement component 8, gamma polypeptide
Gene Aliases
C8C
UniGene
Hs.1285
Gene Chromosome Location
Chr.9: 136944870-136946981 on Build GRCh38
Transcripts
NM_000606.2

SNP Detail

SNP ID Chromosome Location
rs1039610413 Chr.9: 136946849-136946849 on Build GRCh38
rs545894807 Chr.9: 136946772-136946772 on Build GRCh38
rs895606516 Chr.9: 136946881-136946881 on Build GRCh38
rs373036948 Chr.9: 136946807-136946807 on Build GRCh38
rs771007800 Chr.9: 136946773-136946773 on Build GRCh38
rs760080956 Chr.9: 136946827-136946827 on Build GRCh38
rs561880659 Chr.9: 136946861-136946861 on Build GRCh38
rs145014779 Chr.9: 136946786-136946786 on Build GRCh38
rs953506979 Chr.9: 136946963-136946963 on Build GRCh38
rs763408243 Chr.9: 136946790-136946790 on Build GRCh38
rs191866587 Chr.9: 136946862-136946862 on Build GRCh38
rs749987256 Chr.9: 136946821-136946821 on Build GRCh38
rs972073071 Chr.9: 136946957-136946957 on Build GRCh38
rs549740926 Chr.9: 136946922-136946922 on Build GRCh38
rs377687832 Chr.9: 136946785-136946785 on Build GRCh38
rs775954488 Chr.9: 136946805-136946807 on Build GRCh38
rs775529746 Chr.9: 136946788-136946788 on Build GRCh38
rs564153454 Chr.9: 136946900-136946900 on Build GRCh38
rs557797279 Chr.9: 136946971-136946971 on Build GRCh38
rs376631714 Chr.9: 136946815-136946815 on Build GRCh38
rs759967294 Chr.9: 136946946-136946946 on Build GRCh38
rs41309980 Chr.9: 136946760-136946760 on Build GRCh38
rs187101610 Chr.9: 136946828-136946828 on Build GRCh38
rs748141490 Chr.9: 136946841-136946841 on Build GRCh38
rs998173312 Chr.9: 136946863-136946863 on Build GRCh38
rs909757509 Chr.9: 136946833-136946833 on Build GRCh38
rs781320401 Chr.9: 136946784-136946784 on Build GRCh38
rs903255265 Chr.9: 136946895-136946895 on Build GRCh38
rs117447141 Chr.9: 136946917-136946917 on Build GRCh38
rs769075170 Chr.9: 136946795-136946795 on Build GRCh38
rs754803087 Chr.9: 136946832-136946832 on Build GRCh38
rs770094546 Chr.9: 136946772-136946774 on Build GRCh38
rs978745286 Chr.9: 136946920-136946920 on Build GRCh38
rs957569087 Chr.9: 136946916-136946916 on Build GRCh38
rs540895650 Chr.9: 136946896-136946896 on Build GRCh38
rs147752674 Chr.9: 136946848-136946848 on Build GRCh38
rs748008792 Chr.9: 136946761-136946761 on Build GRCh38
rs1051548169 Chr.9: 136946877-136946877 on Build GRCh38
rs556014861 Chr.9: 136946820-136946820 on Build GRCh38
rs1034192118 Chr.9: 136946869-136946869 on Build GRCh38
rs564036334 Chr.9: 136946964-136946965 on Build GRCh38

View Full Product Details