Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33092170-33092443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF3
Entrez Gene ID
1479
Gene Name
cleavage stimulation factor subunit 3
Gene Aliases
CSTF-77
UniGene
Hs.44402
Gene Chromosome Location
Chr.11: 33084584-33161491 on Build GRCh38
Transcripts
NM_001326.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4835976 c.1376-1G>A p.? Chr.11: 33092341-33092341 on Build GRCh38
5640149 c.1435G>A p.E479K Chr.11: 33092281-33092281 on Build GRCh38
3809108 c.1414A>C p.T472P Chr.11: 33092302-33092302 on Build GRCh38
4836262 c.1402G>C p.E468Q Chr.11: 33092314-33092314 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764375258 Chr.11: 33092363-33092364 on Build GRCh38
rs771452356 Chr.11: 33092357-33092357 on Build GRCh38
rs764501503 Chr.11: 33092247-33092247 on Build GRCh38
rs773633893 Chr.11: 33092339-33092339 on Build GRCh38
rs761564137 Chr.11: 33092385-33092385 on Build GRCh38
rs73482844 Chr.11: 33092403-33092403 on Build GRCh38
rs371063770 Chr.11: 33092388-33092388 on Build GRCh38
rs749680289 Chr.11: 33092352-33092352 on Build GRCh38
rs771602234 Chr.11: 33092376-33092376 on Build GRCh38
rs376585351 Chr.11: 33092384-33092384 on Build GRCh38
rs542255754 Chr.11: 33092250-33092250 on Build GRCh38
rs374946238 Chr.11: 33092205-33092205 on Build GRCh38
rs374330250 Chr.11: 33092394-33092394 on Build GRCh38
rs781528275 Chr.11: 33092273-33092273 on Build GRCh38
rs571388005 Chr.11: 33092311-33092311 on Build GRCh38
rs775074543 Chr.11: 33092377-33092377 on Build GRCh38
rs952839299 Chr.11: 33092360-33092360 on Build GRCh38
rs113144162 Chr.11: 33092364-33092365 on Build GRCh38
rs760332671 Chr.11: 33092378-33092378 on Build GRCh38
rs755149209 Chr.11: 33092265-33092265 on Build GRCh38
rs763971493 Chr.11: 33092231-33092231 on Build GRCh38
rs748406781 Chr.11: 33092316-33092316 on Build GRCh38
rs747063822 Chr.11: 33092262-33092262 on Build GRCh38
rs987209279 Chr.11: 33092334-33092334 on Build GRCh38
rs896105222 Chr.11: 33092212-33092212 on Build GRCh38
rs763726992 Chr.11: 33092380-33092380 on Build GRCh38
rs188385129 Chr.11: 33092211-33092211 on Build GRCh38
rs999050451 Chr.11: 33092201-33092201 on Build GRCh38
rs1013595978 Chr.11: 33092300-33092300 on Build GRCh38
rs1023175477 Chr.11: 33092359-33092359 on Build GRCh38
rs769999645 Chr.11: 33092326-33092326 on Build GRCh38
rs757217023 Chr.11: 33092249-33092249 on Build GRCh38
rs71034650 Chr.11: 33092371-33092372 on Build GRCh38
rs386373520 Chr.11: 33092365-33092366 on Build GRCh38
rs562458789 Chr.11: 33092371-33092371 on Build GRCh38
rs961422610 Chr.11: 33092410-33092410 on Build GRCh38
rs774884565 Chr.11: 33092364-33092364 on Build GRCh38

View Full Product Details