Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2237596-2237851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1L2
Entrez Gene ID
1775
Gene Name
deoxyribonuclease I-like 2
Gene Aliases
DNAS1L2
UniGene
Hs.103503
Gene Chromosome Location
Chr.16: 2236252-2238711 on Build GRCh38
Transcripts
NM_001301680.1 NM_001374.2 XM_011522399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6436973 c.594C>A p.D198E Chr.16: 2237769-2237769 on Build GRCh38
1204231 c.673A>G p.S225G Chr.16: 2237848-2237848 on Build GRCh38
3948382 c.576C>G p.D192E Chr.16: 2237634-2237634 on Build GRCh38
6647452 c.634G>A p.V212M Chr.16: 2237809-2237809 on Build GRCh38
4678791 c.591C>T p.D197D Chr.16: 2237649-2237649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755567133 Chr.16: 2237827-2237827 on Build GRCh38
rs200639506 Chr.16: 2237650-2237650 on Build GRCh38
rs772982565 Chr.16: 2237671-2237671 on Build GRCh38
rs779301033 Chr.16: 2237657-2237657 on Build GRCh38
rs767369134 Chr.16: 2237694-2237694 on Build GRCh38
rs8050799 Chr.16: 2237722-2237722 on Build GRCh38
rs769409083 Chr.16: 2237668-2237668 on Build GRCh38
rs779211297 Chr.16: 2237742-2237742 on Build GRCh38
rs62621282 Chr.16: 2237648-2237648 on Build GRCh38
rs780416582 Chr.16: 2237757-2237757 on Build GRCh38
rs776161214 Chr.16: 2237664-2237664 on Build GRCh38
rs572347132 Chr.16: 2237628-2237628 on Build GRCh38
rs773615861 Chr.16: 2237658-2237659 on Build GRCh38
rs922120606 Chr.16: 2237825-2237825 on Build GRCh38
rs762741395 Chr.16: 2237676-2237676 on Build GRCh38
rs1027371135 Chr.16: 2237714-2237714 on Build GRCh38
rs754038870 Chr.16: 2237697-2237697 on Build GRCh38
rs26841 Chr.16: 2237746-2237746 on Build GRCh38
rs750737704 Chr.16: 2237813-2237813 on Build GRCh38
rs756144976 Chr.16: 2237696-2237696 on Build GRCh38
rs752740935 Chr.16: 2237643-2237643 on Build GRCh38
rs376081522 Chr.16: 2237678-2237678 on Build GRCh38
rs747605794 Chr.16: 2237759-2237759 on Build GRCh38
rs757606487 Chr.16: 2237656-2237656 on Build GRCh38
rs746243691 Chr.16: 2237748-2237748 on Build GRCh38
rs751990800 Chr.16: 2237818-2237818 on Build GRCh38
rs753933779 Chr.16: 2237795-2237795 on Build GRCh38
rs1015196900 Chr.16: 2237753-2237753 on Build GRCh38
rs577362963 Chr.16: 2237695-2237695 on Build GRCh38
rs780511185 Chr.16: 2237817-2237817 on Build GRCh38
rs765446336 Chr.16: 2237807-2237807 on Build GRCh38
rs777380182 Chr.16: 2237829-2237829 on Build GRCh38
rs372443542 Chr.16: 2237793-2237793 on Build GRCh38
rs759263300 Chr.16: 2237775-2237775 on Build GRCh38
rs766027556 Chr.16: 2237752-2237760 on Build GRCh38
rs763279413 Chr.16: 2237723-2237745 on Build GRCh38
rs769162157 Chr.16: 2237762-2237762 on Build GRCh38
rs747629349 Chr.16: 2237666-2237666 on Build GRCh38
rs1037513028 Chr.16: 2237808-2237808 on Build GRCh38
rs775284591 Chr.16: 2237783-2237783 on Build GRCh38
rs761980243 Chr.16: 2237796-2237796 on Build GRCh38
rs891692429 Chr.16: 2237674-2237674 on Build GRCh38
rs774317694 Chr.16: 2237686-2237686 on Build GRCh38
rs774222967 Chr.16: 2237773-2237773 on Build GRCh38
rs772886097 Chr.16: 2237766-2237766 on Build GRCh38
rs759467355 Chr.16: 2237688-2237688 on Build GRCh38
rs758886575 Chr.16: 2237755-2237755 on Build GRCh38
rs534305424 Chr.16: 2237629-2237629 on Build GRCh38
rs773907354 Chr.16: 2237801-2237816 on Build GRCh38
rs1001281994 Chr.16: 2237708-2237708 on Build GRCh38
rs758689098 Chr.16: 2237814-2237814 on Build GRCh38
rs760705774 Chr.16: 2237784-2237784 on Build GRCh38
rs767403519 Chr.16: 2237780-2237780 on Build GRCh38
rs1052114476 Chr.16: 2237769-2237769 on Build GRCh38
rs749579347 Chr.16: 2237653-2237653 on Build GRCh38
rs746345357 Chr.16: 2237661-2237661 on Build GRCh38
rs148620653 Chr.16: 2237663-2237663 on Build GRCh38
rs748903104 Chr.16: 2237771-2237771 on Build GRCh38
rs370424080 Chr.16: 2237631-2237631 on Build GRCh38
rs952305130 Chr.16: 2237617-2237617 on Build GRCh38
rs757513865 Chr.16: 2237733-2237733 on Build GRCh38

View Full Product Details