Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109251144-109251389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR2
Entrez Gene ID
1952
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Gene Aliases
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3
UniGene
Hs.57652
Gene Chromosome Location
Chr.1: 109249705-109275756 on Build GRCh38
Transcripts
NM_001408.2 XM_005270580.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5900442 c.1132C>T p.P378S Chr.1: 109251211-109251211 on Build GRCh38
7333761 c.1296C>T p.A432A Chr.1: 109251375-109251375 on Build GRCh38
1294672 c.1265C>T p.T422I Chr.1: 109251344-109251344 on Build GRCh38
7165849 c.1163C>T p.S388F Chr.1: 109251242-109251242 on Build GRCh38
5052363 c.1187delC p.Q398fs*11 Chr.1: 109251266-109251266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369951311 Chr.1: 109251278-109251278 on Build GRCh38
rs755549032 Chr.1: 109251177-109251177 on Build GRCh38
rs193224402 Chr.1: 109251223-109251223 on Build GRCh38
rs769734269 Chr.1: 109251216-109251216 on Build GRCh38
rs375455011 Chr.1: 109251291-109251291 on Build GRCh38
rs149911274 Chr.1: 109251176-109251176 on Build GRCh38
rs199829910 Chr.1: 109251210-109251218 on Build GRCh38
rs748247760 Chr.1: 109251187-109251187 on Build GRCh38
rs573558774 Chr.1: 109251286-109251286 on Build GRCh38
rs768176557 Chr.1: 109251331-109251331 on Build GRCh38
rs373566231 Chr.1: 109251338-109251338 on Build GRCh38
rs754271169 Chr.1: 109251264-109251264 on Build GRCh38
rs367687072 Chr.1: 109251200-109251200 on Build GRCh38
rs944709395 Chr.1: 109251301-109251301 on Build GRCh38
rs550918065 Chr.1: 109251350-109251350 on Build GRCh38
rs149311467 Chr.1: 109251229-109251229 on Build GRCh38
rs775463779 Chr.1: 109251325-109251325 on Build GRCh38
rs747244877 Chr.1: 109251202-109251202 on Build GRCh38
rs201416867 Chr.1: 109251199-109251199 on Build GRCh38
rs567653065 Chr.1: 109251206-109251206 on Build GRCh38
rs759949577 Chr.1: 109251348-109251348 on Build GRCh38
rs780591973 Chr.1: 109251167-109251167 on Build GRCh38
rs773741912 Chr.1: 109251337-109251337 on Build GRCh38
rs759400632 Chr.1: 109251214-109251214 on Build GRCh38
rs780906021 Chr.1: 109251287-109251287 on Build GRCh38
rs148805303 Chr.1: 109251223-109251224 on Build GRCh38
rs926059478 Chr.1: 109251368-109251368 on Build GRCh38
rs200447397 Chr.1: 109251272-109251272 on Build GRCh38
rs142614123 Chr.1: 109251351-109251351 on Build GRCh38
rs747213922 Chr.1: 109251282-109251282 on Build GRCh38
rs755453775 Chr.1: 109251266-109251266 on Build GRCh38
rs758860471 Chr.1: 109251356-109251356 on Build GRCh38
rs202054206 Chr.1: 109251186-109251186 on Build GRCh38
rs769640761 Chr.1: 109251296-109251296 on Build GRCh38
rs776440581 Chr.1: 109251212-109251212 on Build GRCh38
rs1012205619 Chr.1: 109251273-109251273 on Build GRCh38
rs371403541 Chr.1: 109251228-109251228 on Build GRCh38
rs745646802 Chr.1: 109251290-109251290 on Build GRCh38
rs753199359 Chr.1: 109251355-109251355 on Build GRCh38
rs773068309 Chr.1: 109251346-109251346 on Build GRCh38
rs775461482 Chr.1: 109251220-109251220 on Build GRCh38
rs779560542 Chr.1: 109251267-109251267 on Build GRCh38

View Full Product Details