Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 39987857-39988124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK13
Entrez Gene ID
8621
Gene Name
cyclin dependent kinase 13
Gene Aliases
CDC2L, CDC2L5, CHED, hCDK13
UniGene
Hs.233552
Gene Chromosome Location
Chr.7: 39949501-40097134 on Build GRCh38
Transcripts
NM_003718.4 NM_031267.3 XM_011515597.2 XM_017012750.1 XM_017012751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755272 c.1717A>T p.T573S Chr.7: 39988104-39988104 on Build GRCh38
5644430 c.1611delA p.T539fs*7 Chr.7: 39987998-39987998 on Build GRCh38
5660982 c.1617delA p.K540fs*6 Chr.7: 39988004-39988004 on Build GRCh38
150271 c.1498A>G p.T500A Chr.7: 39987885-39987885 on Build GRCh38
107932 c.1625C>T p.P542L Chr.7: 39988012-39988012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150689410 Chr.7: 39988014-39988014 on Build GRCh38
rs771361617 Chr.7: 39987997-39987997 on Build GRCh38
rs776004739 Chr.7: 39988089-39988089 on Build GRCh38
rs112439571 Chr.7: 39987883-39987883 on Build GRCh38
rs138333007 Chr.7: 39988019-39988019 on Build GRCh38
rs747651031 Chr.7: 39987996-39987996 on Build GRCh38
rs746449091 Chr.7: 39988075-39988075 on Build GRCh38
rs764348114 Chr.7: 39988025-39988025 on Build GRCh38
rs761846129 Chr.7: 39987919-39987927 on Build GRCh38
rs750416070 Chr.7: 39988046-39988046 on Build GRCh38
rs376391137 Chr.7: 39988064-39988064 on Build GRCh38
rs759925494 Chr.7: 39988009-39988009 on Build GRCh38
rs778576213 Chr.7: 39988059-39988059 on Build GRCh38
rs867579677 Chr.7: 39987951-39987951 on Build GRCh38
rs767473614 Chr.7: 39988045-39988045 on Build GRCh38
rs763370906 Chr.7: 39988096-39988096 on Build GRCh38
rs781145037 Chr.7: 39987957-39987957 on Build GRCh38
rs200811940 Chr.7: 39988098-39988098 on Build GRCh38
rs753012230 Chr.7: 39987896-39987896 on Build GRCh38
rs751624342 Chr.7: 39988031-39988031 on Build GRCh38
rs772597235 Chr.7: 39988068-39988068 on Build GRCh38
rs145750316 Chr.7: 39987965-39987965 on Build GRCh38
rs768238864 Chr.7: 39987975-39987975 on Build GRCh38
rs1050173183 Chr.7: 39987979-39987979 on Build GRCh38
rs770369397 Chr.7: 39988087-39988087 on Build GRCh38
rs141986436 Chr.7: 39987992-39987992 on Build GRCh38
rs757313115 Chr.7: 39988043-39988043 on Build GRCh38
rs111806120 Chr.7: 39987978-39987978 on Build GRCh38
rs778059575 Chr.7: 39987901-39987901 on Build GRCh38
rs148170327 Chr.7: 39987969-39987969 on Build GRCh38
rs758692200 Chr.7: 39987899-39987899 on Build GRCh38
rs767581253 Chr.7: 39988096-39988098 on Build GRCh38
rs756031403 Chr.7: 39988049-39988049 on Build GRCh38
rs922943045 Chr.7: 39987886-39987886 on Build GRCh38
rs1038941016 Chr.7: 39988017-39988017 on Build GRCh38
rs751601537 Chr.7: 39987929-39987929 on Build GRCh38
rs745673616 Chr.7: 39987958-39987958 on Build GRCh38
rs139716678 Chr.7: 39988058-39988058 on Build GRCh38
rs748996479 Chr.7: 39987973-39987973 on Build GRCh38
rs139418651 Chr.7: 39988012-39988012 on Build GRCh38
rs749014426 Chr.7: 39988057-39988057 on Build GRCh38
rs780672485 Chr.7: 39988047-39988047 on Build GRCh38
rs765670266 Chr.7: 39987888-39987888 on Build GRCh38
rs888833166 Chr.7: 39987999-39987999 on Build GRCh38
rs3735135 Chr.7: 39987885-39987885 on Build GRCh38
rs373649746 Chr.7: 39988063-39988063 on Build GRCh38
rs570306028 Chr.7: 39988011-39988011 on Build GRCh38
rs757148255 Chr.7: 39987945-39987945 on Build GRCh38
rs202207296 Chr.7: 39988051-39988051 on Build GRCh38

View Full Product Details