Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60157641-60157866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA4
Entrez Gene ID
762
Gene Name
carbonic anhydrase 4
Gene Aliases
CAIV, Car4, RP17
UniGene
Hs.89485
Gene Chromosome Location
Chr.17: 60149941-60178118 on Build GRCh38
Transcripts
XM_005257639.2 XM_011525183.2 XM_017025012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792108 c.513+5G>A p.? Chr.17: 60157793-60157793 on Build GRCh38
3520335 c.443G>A p.G148E Chr.17: 60157718-60157718 on Build GRCh38
3520336 c.450G>A p.S150S Chr.17: 60157725-60157725 on Build GRCh38
3196688 c.474C>A p.D158E Chr.17: 60157749-60157749 on Build GRCh38
4549860 c.484G>A p.E162K Chr.17: 60157759-60157759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573405938 Chr.17: 60157759-60157759 on Build GRCh38
rs764137999 Chr.17: 60157680-60157680 on Build GRCh38
rs566521643 Chr.17: 60157832-60157832 on Build GRCh38
rs773737207 Chr.17: 60157721-60157721 on Build GRCh38
rs768606136 Chr.17: 60157696-60157696 on Build GRCh38
rs929390943 Chr.17: 60157681-60157681 on Build GRCh38
rs758158360 Chr.17: 60157750-60157750 on Build GRCh38
rs750247906 Chr.17: 60157746-60157746 on Build GRCh38
rs760076865 Chr.17: 60157663-60157663 on Build GRCh38
rs77550281 Chr.17: 60157813-60157813 on Build GRCh38
rs752763295 Chr.17: 60157836-60157836 on Build GRCh38
rs199805361 Chr.17: 60157686-60157686 on Build GRCh38
rs375432345 Chr.17: 60157806-60157806 on Build GRCh38
rs367637155 Chr.17: 60157722-60157722 on Build GRCh38
rs1002320040 Chr.17: 60157808-60157808 on Build GRCh38
rs200874513 Chr.17: 60157743-60157743 on Build GRCh38
rs139119204 Chr.17: 60157767-60157767 on Build GRCh38
rs933246040 Chr.17: 60157740-60157740 on Build GRCh38
rs778681319 Chr.17: 60157684-60157684 on Build GRCh38
rs751726651 Chr.17: 60157757-60157757 on Build GRCh38
rs749723077 Chr.17: 60157785-60157785 on Build GRCh38
rs150743300 Chr.17: 60157737-60157737 on Build GRCh38
rs751312568 Chr.17: 60157823-60157823 on Build GRCh38
rs753701274 Chr.17: 60157682-60157682 on Build GRCh38
rs981005784 Chr.17: 60157830-60157830 on Build GRCh38
rs762987801 Chr.17: 60157794-60157794 on Build GRCh38
rs766646182 Chr.17: 60157755-60157755 on Build GRCh38
rs748212755 Chr.17: 60157766-60157766 on Build GRCh38
rs748608495 Chr.17: 60157703-60157703 on Build GRCh38
rs992169983 Chr.17: 60157667-60157667 on Build GRCh38
rs774987740 Chr.17: 60157726-60157726 on Build GRCh38
rs549798034 Chr.17: 60157845-60157845 on Build GRCh38
rs763504965 Chr.17: 60157730-60157730 on Build GRCh38
rs747152606 Chr.17: 60157695-60157695 on Build GRCh38
rs531149141 Chr.17: 60157825-60157825 on Build GRCh38
rs760286281 Chr.17: 60157727-60157727 on Build GRCh38
rs778396729 Chr.17: 60157773-60157773 on Build GRCh38
rs781756861 Chr.17: 60157697-60157697 on Build GRCh38
rs781287163 Chr.17: 60157810-60157811 on Build GRCh38
rs185658468 Chr.17: 60157690-60157690 on Build GRCh38
rs766168201 Chr.17: 60157821-60157821 on Build GRCh38
rs779420674 Chr.17: 60157814-60157814 on Build GRCh38
rs919323201 Chr.17: 60157670-60157670 on Build GRCh38
rs200724822 Chr.17: 60157742-60157742 on Build GRCh38
rs550453527 Chr.17: 60157758-60157758 on Build GRCh38
rs746643883 Chr.17: 60157804-60157804 on Build GRCh38
rs368617056 Chr.17: 60157788-60157788 on Build GRCh38
rs896724757 Chr.17: 60157826-60157826 on Build GRCh38
rs762803013 Chr.17: 60157820-60157820 on Build GRCh38
rs201148452 Chr.17: 60157725-60157725 on Build GRCh38
rs757139536 Chr.17: 60157683-60157683 on Build GRCh38
rs755164128 Chr.17: 60157679-60157684 on Build GRCh38
rs201241903 Chr.17: 60157710-60157710 on Build GRCh38
rs371204854 Chr.17: 60157803-60157803 on Build GRCh38
rs867271887 Chr.17: 60157724-60157724 on Build GRCh38

View Full Product Details