Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54999028-54999301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSND
Entrez Gene ID
7809
Gene Name
barttin CLCNK type accessory beta subunit
Gene Aliases
BART, DFNB73
UniGene
Hs.151291
Gene Chromosome Location
Chr.1: 54998944-55008792 on Build GRCh38
Transcripts
NM_057176.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613245 c.88C>T p.R30W Chr.1: 54999274-54999274 on Build GRCh38
1492121 c.74T>C p.L25P Chr.1: 54999260-54999260 on Build GRCh38
7171688 c.78G>A p.M26I Chr.1: 54999264-54999264 on Build GRCh38
6613250 c.58G>T p.A20S Chr.1: 54999244-54999244 on Build GRCh38
6613249 c.22C>T p.R8W Chr.1: 54999208-54999208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145048739 Chr.1: 54999250-54999250 on Build GRCh38
rs769301646 Chr.1: 54999243-54999243 on Build GRCh38
rs144303661 Chr.1: 54999186-54999186 on Build GRCh38
rs759685760 Chr.1: 54999254-54999254 on Build GRCh38
rs748015104 Chr.1: 54999242-54999242 on Build GRCh38
rs74315286 Chr.1: 54999189-54999189 on Build GRCh38
rs141111550 Chr.1: 54999195-54999195 on Build GRCh38
rs373288770 Chr.1: 54999144-54999144 on Build GRCh38
rs754403589 Chr.1: 54999238-54999238 on Build GRCh38
rs564571762 Chr.1: 54999251-54999251 on Build GRCh38
rs778929707 Chr.1: 54999241-54999241 on Build GRCh38
rs74315287 Chr.1: 54999214-54999214 on Build GRCh38
rs567238039 Chr.1: 54999277-54999277 on Build GRCh38
rs371937424 Chr.1: 54999213-54999213 on Build GRCh38
rs1048152752 Chr.1: 54999159-54999159 on Build GRCh38
rs373918982 Chr.1: 54999143-54999143 on Build GRCh38
rs768683733 Chr.1: 54999153-54999153 on Build GRCh38
rs765805682 Chr.1: 54999203-54999203 on Build GRCh38
rs781012759 Chr.1: 54999142-54999142 on Build GRCh38
rs752014769 Chr.1: 54999273-54999273 on Build GRCh38
rs201119972 Chr.1: 54999275-54999275 on Build GRCh38
rs188418228 Chr.1: 54999162-54999162 on Build GRCh38
rs751967664 Chr.1: 54999217-54999217 on Build GRCh38
rs2500341 Chr.1: 54999117-54999117 on Build GRCh38
rs763620250 Chr.1: 54999261-54999261 on Build GRCh38
rs368632305 Chr.1: 54999255-54999255 on Build GRCh38
rs376897764 Chr.1: 54999152-54999152 on Build GRCh38
rs74315284 Chr.1: 54999187-54999187 on Build GRCh38
rs779547279 Chr.1: 54999222-54999222 on Build GRCh38
rs758796444 Chr.1: 54999141-54999141 on Build GRCh38
rs74315288 Chr.1: 54999209-54999209 on Build GRCh38
rs121908145 Chr.1: 54999196-54999196 on Build GRCh38
rs199729521 Chr.1: 54999225-54999225 on Build GRCh38
rs778681727 Chr.1: 54999157-54999157 on Build GRCh38
rs776067180 Chr.1: 54999258-54999258 on Build GRCh38
rs74315285 Chr.1: 54999208-54999208 on Build GRCh38
rs2500340 Chr.1: 54999070-54999070 on Build GRCh38
rs112179763 Chr.1: 54999104-54999104 on Build GRCh38
rs779777664 Chr.1: 54999145-54999145 on Build GRCh38
rs990973036 Chr.1: 54999118-54999118 on Build GRCh38
rs201342416 Chr.1: 54999202-54999202 on Build GRCh38
rs762233340 Chr.1: 54999274-54999274 on Build GRCh38
rs575695982 Chr.1: 54999109-54999109 on Build GRCh38
rs942566338 Chr.1: 54999172-54999172 on Build GRCh38
rs764685237 Chr.1: 54999269-54999269 on Build GRCh38
rs558960277 Chr.1: 54999071-54999071 on Build GRCh38
rs770422666 Chr.1: 54999203-54999203 on Build GRCh38
rs121908144 Chr.1: 54999221-54999221 on Build GRCh38
rs141611486 Chr.1: 54999249-54999249 on Build GRCh38

View Full Product Details