Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163441710-163441965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG1
Entrez Gene ID
900
Gene Name
cyclin G1
Gene Aliases
CCNG
UniGene
Hs.79101
Gene Chromosome Location
Chr.5: 163437567-163445016 on Build GRCh38
Transcripts
NM_004060.3 NM_199246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413763 c.559A>T p.K187* Chr.5: 163441926-163441926 on Build GRCh38
3697118 c.532A>C p.N178H Chr.5: 163441899-163441899 on Build GRCh38
1065364 c.563C>T p.A188V Chr.5: 163441930-163441930 on Build GRCh38
7360108 c.537T>G p.F179L Chr.5: 163441904-163441904 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs907068894 Chr.5: 163441751-163441751 on Build GRCh38
rs751919236 Chr.5: 163441847-163441847 on Build GRCh38
rs2069352 Chr.5: 163441899-163441899 on Build GRCh38
rs772517082 Chr.5: 163441836-163441836 on Build GRCh38
rs753265525 Chr.5: 163441860-163441860 on Build GRCh38
rs1803357 Chr.5: 163441888-163441888 on Build GRCh38
rs552028981 Chr.5: 163441830-163441830 on Build GRCh38
rs771905774 Chr.5: 163441932-163441932 on Build GRCh38
rs776129993 Chr.5: 163441871-163441872 on Build GRCh38
rs776051149 Chr.5: 163441818-163441818 on Build GRCh38
rs938591075 Chr.5: 163441790-163441790 on Build GRCh38
rs11541970 Chr.5: 163441904-163441904 on Build GRCh38
rs186641024 Chr.5: 163441813-163441813 on Build GRCh38
rs925775746 Chr.5: 163441758-163441758 on Build GRCh38
rs778754070 Chr.5: 163441912-163441912 on Build GRCh38
rs373510789 Chr.5: 163441913-163441913 on Build GRCh38
rs368421496 Chr.5: 163441849-163441849 on Build GRCh38
rs370703875 Chr.5: 163441925-163441925 on Build GRCh38
rs755499407 Chr.5: 163441848-163441848 on Build GRCh38
rs1011539543 Chr.5: 163441852-163441852 on Build GRCh38
rs755631512 Chr.5: 163441861-163441861 on Build GRCh38

View Full Product Details