Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41158709-41158934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6
Entrez Gene ID
729
Gene Name
complement component 6
Gene Aliases
-
UniGene
Hs.481992
Gene Chromosome Location
Chr.5: 41142116-41261486 on Build GRCh38
Transcripts
NM_000065.3 NM_001115131.2 XM_005248357.2 XM_006714496.3 XM_011514114.2 XM_011514115.2 XM_011514116.2 XM_011514117.2 XM_011514118.2 XM_011514119.2 XM_011514121.2 XM_017009818.1 XM_017009819.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7159028 c.1908T>A p.P636P Chr.5: 41158734-41158734 on Build GRCh38
4594733 c.1881C>T p.D627D Chr.5: 41158761-41158761 on Build GRCh38
1437609 c.1876G>T p.D626Y Chr.5: 41158766-41158766 on Build GRCh38
3941323 c.1901A>G p.D634G Chr.5: 41158741-41158741 on Build GRCh38
4933360 c.1875T>G p.N625K Chr.5: 41158767-41158767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750486659 Chr.5: 41158777-41158777 on Build GRCh38
rs548480440 Chr.5: 41158814-41158814 on Build GRCh38
rs778114536 Chr.5: 41158909-41158912 on Build GRCh38
rs140790579 Chr.5: 41158865-41158865 on Build GRCh38
rs774130582 Chr.5: 41158837-41158837 on Build GRCh38
rs1032386564 Chr.5: 41158842-41158842 on Build GRCh38
rs368142485 Chr.5: 41158734-41158734 on Build GRCh38
rs779359523 Chr.5: 41158811-41158811 on Build GRCh38
rs769453887 Chr.5: 41158743-41158743 on Build GRCh38
rs764785820 Chr.5: 41158820-41158835 on Build GRCh38
rs767651698 Chr.5: 41158772-41158772 on Build GRCh38
rs188723807 Chr.5: 41158830-41158830 on Build GRCh38
rs780345600 Chr.5: 41158781-41158781 on Build GRCh38
rs576436354 Chr.5: 41158803-41158803 on Build GRCh38
rs183319289 Chr.5: 41158820-41158820 on Build GRCh38
rs751514430 Chr.5: 41158764-41158764 on Build GRCh38
rs146901928 Chr.5: 41158854-41158854 on Build GRCh38
rs544678725 Chr.5: 41158742-41158742 on Build GRCh38
rs957832609 Chr.5: 41158864-41158864 on Build GRCh38
rs201273657 Chr.5: 41158739-41158739 on Build GRCh38
rs866261314 Chr.5: 41158752-41158752 on Build GRCh38
rs186960114 Chr.5: 41158736-41158736 on Build GRCh38
rs761471336 Chr.5: 41158814-41158817 on Build GRCh38
rs76410478 Chr.5: 41158807-41158807 on Build GRCh38
rs913642567 Chr.5: 41158870-41158870 on Build GRCh38
rs372345589 Chr.5: 41158824-41158824 on Build GRCh38
rs61469168 Chr.5: 41158763-41158763 on Build GRCh38
rs6866352 Chr.5: 41158761-41158761 on Build GRCh38
rs780886156 Chr.5: 41158816-41158816 on Build GRCh38
rs762841413 Chr.5: 41158753-41158753 on Build GRCh38
rs41271069 Chr.5: 41158879-41158879 on Build GRCh38
rs41271071 Chr.5: 41158884-41158884 on Build GRCh38
rs756339343 Chr.5: 41158778-41158778 on Build GRCh38
rs193022672 Chr.5: 41158889-41158889 on Build GRCh38
rs375900177 Chr.5: 41158828-41158828 on Build GRCh38
rs904769290 Chr.5: 41158794-41158794 on Build GRCh38
rs745409150 Chr.5: 41158817-41158817 on Build GRCh38
rs776264890 Chr.5: 41158738-41158738 on Build GRCh38
rs774959389 Chr.5: 41158748-41158748 on Build GRCh38

View Full Product Details