Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52355683-52355908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1B
Entrez Gene ID
200014
Gene Name
coiled-coil and C2 domain containing 1B
Gene Aliases
-
UniGene
Hs.591451
Gene Chromosome Location
Chr.1: 52348437-52366291 on Build GRCh38
Transcripts
NM_032449.2 XM_005270590.2 XM_005270591.2 XM_005270593.2 XM_006710423.2 XM_017000559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1185465 c.2103G>C p.M701I Chr.1: 52355814-52355814 on Build GRCh38
5646136 c.208G>A p.E70K Chr.1: 52355834-52355834 on Build GRCh38
5646135 c.2083G>A p.E695K Chr.1: 52355834-52355834 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs552055830 Chr.1: 52355709-52355709 on Build GRCh38
rs745406819 Chr.1: 52355829-52355829 on Build GRCh38
rs754052636 Chr.1: 52355806-52355806 on Build GRCh38
rs748933095 Chr.1: 52355851-52355851 on Build GRCh38
rs748719065 Chr.1: 52355814-52355814 on Build GRCh38
rs745548813 Chr.1: 52355774-52355774 on Build GRCh38
rs762752207 Chr.1: 52355780-52355780 on Build GRCh38
rs750464972 Chr.1: 52355786-52355786 on Build GRCh38
rs148278563 Chr.1: 52355805-52355805 on Build GRCh38
rs750480906 Chr.1: 52355798-52355798 on Build GRCh38
rs567841906 Chr.1: 52355823-52355823 on Build GRCh38
rs371310320 Chr.1: 52355740-52355740 on Build GRCh38
rs755343361 Chr.1: 52355751-52355751 on Build GRCh38
rs761269563 Chr.1: 52355757-52355757 on Build GRCh38
rs770409354 Chr.1: 52355757-52355757 on Build GRCh38
rs555821949 Chr.1: 52355813-52355813 on Build GRCh38
rs35495854 Chr.1: 52355784-52355785 on Build GRCh38
rs751442751 Chr.1: 52355723-52355723 on Build GRCh38
rs763808717 Chr.1: 52355781-52355781 on Build GRCh38
rs1031965686 Chr.1: 52355714-52355714 on Build GRCh38
rs758911649 Chr.1: 52355816-52355816 on Build GRCh38
rs766643268 Chr.1: 52355803-52355803 on Build GRCh38
rs779570771 Chr.1: 52355752-52355752 on Build GRCh38
rs372864749 Chr.1: 52355728-52355728 on Build GRCh38
rs190650217 Chr.1: 52355706-52355706 on Build GRCh38
rs374619861 Chr.1: 52355857-52355857 on Build GRCh38
rs757332239 Chr.1: 52355725-52355725 on Build GRCh38
rs956699699 Chr.1: 52355721-52355722 on Build GRCh38
rs772970705 Chr.1: 52355886-52355886 on Build GRCh38
rs374468381 Chr.1: 52355730-52355730 on Build GRCh38
rs769129835 Chr.1: 52355842-52355842 on Build GRCh38
rs981375203 Chr.1: 52355767-52355767 on Build GRCh38
rs951513546 Chr.1: 52355825-52355825 on Build GRCh38
rs150767179 Chr.1: 52355812-52355812 on Build GRCh38
rs769517590 Chr.1: 52355776-52355776 on Build GRCh38
rs781769185 Chr.1: 52355756-52355756 on Build GRCh38
rs756102445 Chr.1: 52355802-52355802 on Build GRCh38
rs928143826 Chr.1: 52355773-52355775 on Build GRCh38
rs774110312 Chr.1: 52355856-52355856 on Build GRCh38
rs939636188 Chr.1: 52355800-52355800 on Build GRCh38
rs775129825 Chr.1: 52355847-52355847 on Build GRCh38
rs772075326 Chr.1: 52355858-52355858 on Build GRCh38
rs367732982 Chr.1: 52355731-52355731 on Build GRCh38
rs767628463 Chr.1: 52355727-52355727 on Build GRCh38
rs761724893 Chr.1: 52355796-52355796 on Build GRCh38
rs768496404 Chr.1: 52355852-52355852 on Build GRCh38
rs776185734 Chr.1: 52355771-52355771 on Build GRCh38
rs774337211 Chr.1: 52355792-52355792 on Build GRCh38
rs767542806 Chr.1: 52355797-52355797 on Build GRCh38

View Full Product Details