Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103521326-103521575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKB
Entrez Gene ID
1152
Gene Name
creatine kinase B
Gene Aliases
B-CK, BCK, CKBB, HEL-211, HEL-S-29
UniGene
Hs.173724
Gene Chromosome Location
Chr.14: 103519658-103522859 on Build GRCh38
Transcripts
NM_001823.4 XM_017020951.1 XM_017020952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629592 c.554A>T p.Q185L Chr.14: 103521362-103521362 on Build GRCh38
6053564 c.532A>T p.S178C Chr.14: 103521384-103521384 on Build GRCh38
4136234 c.498C>G p.D166E Chr.14: 103521418-103521418 on Build GRCh38
6426509 c.494T>C p.L165P Chr.14: 103521422-103521422 on Build GRCh38
7180659 c.568G>C p.D190H Chr.14: 103521348-103521348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763157009 Chr.14: 103521466-103521466 on Build GRCh38
rs774233084 Chr.14: 103521438-103521438 on Build GRCh38
rs34995931 Chr.14: 103521544-103521544 on Build GRCh38
rs756873696 Chr.14: 103521480-103521480 on Build GRCh38
rs768646865 Chr.14: 103521424-103521424 on Build GRCh38
rs1037618915 Chr.14: 103521368-103521368 on Build GRCh38
rs200890896 Chr.14: 103521391-103521391 on Build GRCh38
rs544177157 Chr.14: 103521507-103521507 on Build GRCh38
rs983535914 Chr.14: 103521559-103521559 on Build GRCh38
rs1041105054 Chr.14: 103521351-103521351 on Build GRCh38
rs968552046 Chr.14: 103521494-103521494 on Build GRCh38
rs374321801 Chr.14: 103521476-103521476 on Build GRCh38
rs754012287 Chr.14: 103521348-103521348 on Build GRCh38
rs769546303 Chr.14: 103521407-103521407 on Build GRCh38
rs759797744 Chr.14: 103521444-103521444 on Build GRCh38
rs778366070 Chr.14: 103521387-103521387 on Build GRCh38
rs759868311 Chr.14: 103521367-103521367 on Build GRCh38
rs764724502 Chr.14: 103521470-103521470 on Build GRCh38
rs756879188 Chr.14: 103521467-103521468 on Build GRCh38
rs750103975 Chr.14: 103521484-103521484 on Build GRCh38
rs760766301 Chr.14: 103521439-103521439 on Build GRCh38
rs36002620 Chr.14: 103521386-103521386 on Build GRCh38
rs776880074 Chr.14: 103521443-103521443 on Build GRCh38
rs770978803 Chr.14: 103521441-103521441 on Build GRCh38
rs753527319 Chr.14: 103521416-103521416 on Build GRCh38
rs369278146 Chr.14: 103521485-103521485 on Build GRCh38
rs911012665 Chr.14: 103521503-103521503 on Build GRCh38
rs767133418 Chr.14: 103521481-103521481 on Build GRCh38
rs1035441563 Chr.14: 103521475-103521475 on Build GRCh38
rs781011154 Chr.14: 103521393-103521393 on Build GRCh38
rs765604703 Chr.14: 103521381-103521381 on Build GRCh38
rs749139920 Chr.14: 103521486-103521486 on Build GRCh38
rs753209091 Chr.14: 103521382-103521382 on Build GRCh38
rs888633414 Chr.14: 103521409-103521409 on Build GRCh38
rs558681211 Chr.14: 103521498-103521498 on Build GRCh38
rs575158421 Chr.14: 103521505-103521505 on Build GRCh38
rs745508721 Chr.14: 103521402-103521402 on Build GRCh38
rs1023746346 Chr.14: 103521454-103521454 on Build GRCh38
rs753117577 Chr.14: 103521462-103521462 on Build GRCh38
rs202115891 Chr.14: 103521460-103521460 on Build GRCh38
rs1047304467 Chr.14: 103521392-103521392 on Build GRCh38
rs928652100 Chr.14: 103521528-103521528 on Build GRCh38
rs200266751 Chr.14: 103521417-103521417 on Build GRCh38

View Full Product Details