Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 62771210-62771481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHBP1
Entrez Gene ID
23301
Gene Name
EH domain binding protein 1
Gene Aliases
HPC12, NACSIN
UniGene
Hs.271667
Gene Chromosome Location
Chr.2: 62673851-63046487 on Build GRCh38
Transcripts
NM_001142614.1 NM_001142615.2 NM_001142616.1 NM_015252.3 XM_005264225.2 XM_005264226.2 XM_005264227.1 XM_005264228.2 XM_006711971.2 XM_011532713.2 XM_011532714.2 XM_011532715.2 XM_011532716.2 XM_011532717.1 XM_011532718.2 XM_017003643.1 XM_017003644.1 XM_017003645.1 XM_017003646.1 XM_017003647.1 XM_017003648.1 XM_017003649.1 XM_017003650.1 XM_017003651.1 XM_017003652.1 XM_017003653.1 XM_017003654.1 XM_017003655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4917532 c.308A>C p.E103A Chr.2: 62771388-62771388 on Build GRCh38
359449 c.269C>T p.A90V Chr.2: 62771349-62771349 on Build GRCh38
4539683 c.271G>A p.E91K Chr.2: 62771351-62771351 on Build GRCh38
3391807 c.259-3C>T p.? Chr.2: 62771336-62771336 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375051545 Chr.2: 62771307-62771307 on Build GRCh38
rs201855702 Chr.2: 62771400-62771400 on Build GRCh38
rs866700227 Chr.2: 62771376-62771376 on Build GRCh38
rs763676324 Chr.2: 62771300-62771300 on Build GRCh38
rs758671591 Chr.2: 62771384-62771384 on Build GRCh38
rs761348668 Chr.2: 62771429-62771429 on Build GRCh38
rs916927601 Chr.2: 62771257-62771257 on Build GRCh38
rs1009568329 Chr.2: 62771259-62771259 on Build GRCh38
rs766781594 Chr.2: 62771339-62771339 on Build GRCh38
rs748332929 Chr.2: 62771377-62771377 on Build GRCh38
rs1053860015 Chr.2: 62771242-62771242 on Build GRCh38
rs779080782 Chr.2: 62771353-62771353 on Build GRCh38
rs764938164 Chr.2: 62771319-62771319 on Build GRCh38
rs760889210 Chr.2: 62771439-62771439 on Build GRCh38
rs768015523 Chr.2: 62771423-62771423 on Build GRCh38
rs1054747926 Chr.2: 62771336-62771336 on Build GRCh38
rs892589759 Chr.2: 62771258-62771258 on Build GRCh38
rs373948377 Chr.2: 62771418-62771418 on Build GRCh38
rs577156978 Chr.2: 62771281-62771281 on Build GRCh38
rs772628096 Chr.2: 62771436-62771437 on Build GRCh38
rs766769630 Chr.2: 62771450-62771450 on Build GRCh38
rs75835972 Chr.2: 62771350-62771350 on Build GRCh38
rs756565681 Chr.2: 62771328-62771328 on Build GRCh38
rs762022722 Chr.2: 62771312-62771312 on Build GRCh38
rs745656110 Chr.2: 62771398-62771398 on Build GRCh38
rs767149214 Chr.2: 62771437-62771437 on Build GRCh38
rs200927057 Chr.2: 62771349-62771349 on Build GRCh38
rs1047411092 Chr.2: 62771237-62771237 on Build GRCh38
rs933017912 Chr.2: 62771417-62771417 on Build GRCh38
rs750753417 Chr.2: 62771324-62771324 on Build GRCh38
rs772904659 Chr.2: 62771438-62771438 on Build GRCh38
rs922901399 Chr.2: 62771413-62771413 on Build GRCh38
rs548907699 Chr.2: 62771424-62771424 on Build GRCh38
rs762361231 Chr.2: 62771297-62771297 on Build GRCh38
rs958515710 Chr.2: 62771332-62771332 on Build GRCh38
rs1044202792 Chr.2: 62771289-62771289 on Build GRCh38
rs369784262 Chr.2: 62771411-62771411 on Build GRCh38
rs369211113 Chr.2: 62771238-62771238 on Build GRCh38
rs778182203 Chr.2: 62771392-62771392 on Build GRCh38
rs961729487 Chr.2: 62771370-62771370 on Build GRCh38
rs773929122 Chr.2: 62771301-62771301 on Build GRCh38

View Full Product Details