Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17826009-17826248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL8
Entrez Gene ID
81569
Gene Name
actin like 8
Gene Aliases
CT57
UniGene
Hs.2149
Gene Chromosome Location
Chr.1: 17755313-17827063 on Build GRCh38
Transcripts
NM_030812.2 XM_011542212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2096452 c.673C>T p.R225C Chr.1: 17826091-17826091 on Build GRCh38
6592933 c.817C>T p.P273S Chr.1: 17826235-17826235 on Build GRCh38
5511463 c.734G>A p.R245H Chr.1: 17826152-17826152 on Build GRCh38
6592935 c.802C>T p.P268S Chr.1: 17826220-17826220 on Build GRCh38
5526210 c.814A>G p.I272V Chr.1: 17826232-17826232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142430342 Chr.1: 17826035-17826035 on Build GRCh38
rs773088483 Chr.1: 17826167-17826167 on Build GRCh38
rs374422437 Chr.1: 17826190-17826190 on Build GRCh38
rs776381938 Chr.1: 17826034-17826034 on Build GRCh38
rs560722176 Chr.1: 17826055-17826055 on Build GRCh38
rs764628198 Chr.1: 17826139-17826139 on Build GRCh38
rs369481049 Chr.1: 17826090-17826090 on Build GRCh38
rs771910306 Chr.1: 17826030-17826030 on Build GRCh38
rs267598220 Chr.1: 17826091-17826091 on Build GRCh38
rs754579549 Chr.1: 17826222-17826222 on Build GRCh38
rs760320099 Chr.1: 17826033-17826033 on Build GRCh38
rs532283065 Chr.1: 17826169-17826169 on Build GRCh38
rs765781583 Chr.1: 17826220-17826220 on Build GRCh38
rs762708237 Chr.1: 17826057-17826057 on Build GRCh38
rs148049229 Chr.1: 17826154-17826154 on Build GRCh38
rs765268296 Chr.1: 17826048-17826048 on Build GRCh38
rs751796228 Chr.1: 17826176-17826176 on Build GRCh38
rs3795322 Chr.1: 17826151-17826151 on Build GRCh38
rs780827812 Chr.1: 17826223-17826223 on Build GRCh38
rs753114996 Chr.1: 17826070-17826070 on Build GRCh38
rs745438404 Chr.1: 17826074-17826074 on Build GRCh38
rs201229770 Chr.1: 17826072-17826072 on Build GRCh38
rs748011406 Chr.1: 17826082-17826082 on Build GRCh38
rs746694408 Chr.1: 17826079-17826079 on Build GRCh38
rs779794365 Chr.1: 17826076-17826076 on Build GRCh38
rs781177786 Chr.1: 17826175-17826176 on Build GRCh38
rs1030239557 Chr.1: 17826042-17826042 on Build GRCh38
rs778553946 Chr.1: 17826073-17826073 on Build GRCh38
rs371142860 Chr.1: 17826063-17826063 on Build GRCh38
rs148875551 Chr.1: 17826152-17826152 on Build GRCh38
rs982031545 Chr.1: 17826226-17826226 on Build GRCh38
rs754355183 Chr.1: 17826204-17826204 on Build GRCh38
rs756685710 Chr.1: 17826132-17826132 on Build GRCh38
rs754410089 Chr.1: 17826144-17826144 on Build GRCh38
rs145930628 Chr.1: 17826170-17826170 on Build GRCh38
rs478698 Chr.1: 17826087-17826087 on Build GRCh38
rs970469065 Chr.1: 17826225-17826225 on Build GRCh38
rs757878765 Chr.1: 17826213-17826213 on Build GRCh38
rs768293838 Chr.1: 17826080-17826080 on Build GRCh38
rs757579662 Chr.1: 17826120-17826120 on Build GRCh38
rs985809409 Chr.1: 17826059-17826059 on Build GRCh38
rs751134742 Chr.1: 17826221-17826221 on Build GRCh38
rs755252726 Chr.1: 17826065-17826065 on Build GRCh38
rs1040201800 Chr.1: 17826138-17826138 on Build GRCh38
rs769593730 Chr.1: 17826166-17826166 on Build GRCh38
rs781646116 Chr.1: 17826067-17826067 on Build GRCh38
rs146517291 Chr.1: 17826102-17826102 on Build GRCh38
rs748039028 Chr.1: 17826165-17826165 on Build GRCh38
rs372611534 Chr.1: 17826031-17826031 on Build GRCh38
rs761062155 Chr.1: 17826178-17826178 on Build GRCh38
rs747053631 Chr.1: 17826103-17826104 on Build GRCh38
rs202116436 Chr.1: 17826200-17826200 on Build GRCh38
rs769777106 Chr.1: 17826083-17826083 on Build GRCh38
rs746607498 Chr.1: 17826153-17826153 on Build GRCh38
rs759666029 Chr.1: 17826174-17826174 on Build GRCh38
rs759844551 Chr.1: 17826096-17826096 on Build GRCh38
rs141563095 Chr.1: 17826214-17826214 on Build GRCh38
rs780831696 Chr.1: 17826164-17826164 on Build GRCh38
rs376409500 Chr.1: 17826062-17826062 on Build GRCh38
rs771041754 Chr.1: 17826139-17826143 on Build GRCh38

View Full Product Details