Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60265904-60266149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA8
Entrez Gene ID
767
Gene Name
carbonic anhydrase 8
Gene Aliases
CA-RP, CA-VIII, CALS, CAMRQ3, CARP
UniGene
Hs.654388
Gene Chromosome Location
Chr.8: 60185420-60281423 on Build GRCh38
Transcripts
NM_001321837.1 NM_001321838.1 NM_001321839.1 NM_004056.5 NR_135821.1 XM_011517587.2 XM_011517588.2 XM_017013818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100756 c.340G>A p.E114K Chr.8: 60266002-60266002 on Build GRCh38
6113630 c.412A>G p.M138V Chr.8: 60265930-60265930 on Build GRCh38
5483279 c.417G>C p.E139D Chr.8: 60265925-60265925 on Build GRCh38
4544830 c.364G>A p.E122K Chr.8: 60265978-60265978 on Build GRCh38
3763364 c.327A>G p.E109E Chr.8: 60266015-60266015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368293349 Chr.8: 60266025-60266025 on Build GRCh38
rs768734920 Chr.8: 60266073-60266073 on Build GRCh38
rs73685412 Chr.8: 60266033-60266033 on Build GRCh38
rs539466597 Chr.8: 60266118-60266119 on Build GRCh38
rs149391728 Chr.8: 60266043-60266043 on Build GRCh38
rs576481192 Chr.8: 60266119-60266120 on Build GRCh38
rs747934440 Chr.8: 60266019-60266019 on Build GRCh38
rs371686661 Chr.8: 60266074-60266074 on Build GRCh38
rs553771087 Chr.8: 60265980-60265980 on Build GRCh38
rs754202196 Chr.8: 60265939-60265939 on Build GRCh38
rs146024789 Chr.8: 60265952-60265952 on Build GRCh38
rs906750876 Chr.8: 60265969-60265969 on Build GRCh38
rs113930494 Chr.8: 60266113-60266113 on Build GRCh38
rs775697179 Chr.8: 60265934-60265934 on Build GRCh38
rs923906967 Chr.8: 60266092-60266092 on Build GRCh38
rs751282028 Chr.8: 60266007-60266007 on Build GRCh38
rs543917887 Chr.8: 60266065-60266065 on Build GRCh38
rs762193358 Chr.8: 60266080-60266080 on Build GRCh38
rs201131573 Chr.8: 60266002-60266002 on Build GRCh38
rs62951562 Chr.8: 60266128-60266128 on Build GRCh38
rs796551074 Chr.8: 60266128-60266128 on Build GRCh38
rs749173414 Chr.8: 60266042-60266042 on Build GRCh38
rs5891756 Chr.8: 60266118-60266118 on Build GRCh38
rs200882621 Chr.8: 60265968-60265968 on Build GRCh38
rs761028392 Chr.8: 60266072-60266072 on Build GRCh38
rs267606695 Chr.8: 60266044-60266044 on Build GRCh38
rs557692285 Chr.8: 60266119-60266119 on Build GRCh38
rs984336736 Chr.8: 60266016-60266016 on Build GRCh38
rs935140945 Chr.8: 60266105-60266105 on Build GRCh38
rs139958315 Chr.8: 60265953-60265953 on Build GRCh38
rs773720367 Chr.8: 60266089-60266089 on Build GRCh38
rs779371302 Chr.8: 60266003-60266003 on Build GRCh38
rs61743130 Chr.8: 60266008-60266008 on Build GRCh38
rs7464181 Chr.8: 60266015-60266015 on Build GRCh38
rs765516429 Chr.8: 60266087-60266087 on Build GRCh38
rs975357204 Chr.8: 60266063-60266063 on Build GRCh38

View Full Product Details