Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65870335-65870600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFEMP2
Entrez Gene ID
30008
Gene Name
EGF containing fibulin like extracellular matrix protein 2
Gene Aliases
ARCL1B, FBLN4, MBP1, UPH1
UniGene
Hs.731454
Gene Chromosome Location
Chr.11: 65866441-65872934 on Build GRCh38
Transcripts
NM_016938.4 NR_037718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
930531 c.474C>T p.I158I Chr.11: 65870552-65870552 on Build GRCh38
3451912 c.483G>A p.E161E Chr.11: 65870543-65870543 on Build GRCh38
1605043 c.490G>T p.D164Y Chr.11: 65870536-65870536 on Build GRCh38
6651967 c.475G>A p.G159R Chr.11: 65870551-65870551 on Build GRCh38
1204983 c.463T>C p.Y155H Chr.11: 65870563-65870563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767599210 Chr.11: 65870546-65870546 on Build GRCh38
rs630394 Chr.11: 65870513-65870513 on Build GRCh38
rs749512099 Chr.11: 65870392-65870392 on Build GRCh38
rs1001580366 Chr.11: 65870522-65870522 on Build GRCh38
rs372863827 Chr.11: 65870544-65870544 on Build GRCh38
rs942430661 Chr.11: 65870441-65870441 on Build GRCh38
rs1018063204 Chr.11: 65870417-65870417 on Build GRCh38
rs377199919 Chr.11: 65870566-65870566 on Build GRCh38
rs754676480 Chr.11: 65870555-65870555 on Build GRCh38
rs200069197 Chr.11: 65870494-65870494 on Build GRCh38
rs143247661 Chr.11: 65870571-65870571 on Build GRCh38
rs765318078 Chr.11: 65870540-65870540 on Build GRCh38
rs2234463 Chr.11: 65870375-65870375 on Build GRCh38
rs111927255 Chr.11: 65870416-65870416 on Build GRCh38
rs72924781 Chr.11: 65870448-65870448 on Build GRCh38
rs891781482 Chr.11: 65870485-65870485 on Build GRCh38
rs779614307 Chr.11: 65870552-65870552 on Build GRCh38
rs761656636 Chr.11: 65870545-65870545 on Build GRCh38
rs573771187 Chr.11: 65870560-65870560 on Build GRCh38
rs892934459 Chr.11: 65870421-65870421 on Build GRCh38
rs967339865 Chr.11: 65870460-65870460 on Build GRCh38
rs935260345 Chr.11: 65870462-65870462 on Build GRCh38
rs1052806457 Chr.11: 65870483-65870483 on Build GRCh38
rs569328716 Chr.11: 65870439-65870439 on Build GRCh38
rs1051793199 Chr.11: 65870391-65870391 on Build GRCh38
rs778303057 Chr.11: 65870563-65870563 on Build GRCh38
rs759807856 Chr.11: 65870525-65870525 on Build GRCh38
rs981496832 Chr.11: 65870492-65870492 on Build GRCh38
rs553867624 Chr.11: 65870547-65870547 on Build GRCh38
rs760786325 Chr.11: 65870505-65870505 on Build GRCh38
rs755635112 Chr.11: 65870550-65870550 on Build GRCh38
rs1043545284 Chr.11: 65870445-65870445 on Build GRCh38
rs374498682 Chr.11: 65870506-65870506 on Build GRCh38
rs778385325 Chr.11: 65870559-65870559 on Build GRCh38

View Full Product Details