Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77901099-77901370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDNRB
Entrez Gene ID
1910
Gene Name
endothelin receptor type B
Gene Aliases
ABCDS, ET-B, ET-BR, ETB, ETB1, ETBR, ETRB, HSCR, HSCR2, WS4A
UniGene
Hs.732046
Gene Chromosome Location
Chr.13: 77895481-77975529 on Build GRCh38
Transcripts
NM_000115.3 NM_001122659.2 NM_001201397.1 NM_003991.3 NR_103853.1

Gene Symbol
EDNRB-AS1
Entrez Gene ID
100505518
Gene Name
EDNRB antisense RNA 1
Gene Aliases
-
UniGene
Hs.742215
Gene Chromosome Location
Chr.13: 77818937-77908442 on Build GRCh38
Transcripts
NM_000115.3 NM_001122659.2 NM_001201397.1 NM_003991.3 NR_103853.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6139579 c.893G>T p.C298F Chr.13: 77901116-77901116 on Build GRCh38
5903047 c.820G>A p.D274N Chr.13: 77901189-77901189 on Build GRCh38
6277132 c.802-8C>T p.? Chr.13: 77901215-77901215 on Build GRCh38
3753749 c.831A>G p.L277L Chr.13: 77901178-77901178 on Build GRCh38
3469825 c.884T>C p.L295P Chr.13: 77901125-77901125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757371484 Chr.13: 77901139-77901139 on Build GRCh38
rs771549175 Chr.13: 77901201-77901201 on Build GRCh38
rs746051328 Chr.13: 77901173-77901173 on Build GRCh38
rs1010877652 Chr.13: 77901313-77901313 on Build GRCh38
rs543375862 Chr.13: 77901133-77901133 on Build GRCh38
rs113657418 Chr.13: 77901168-77901168 on Build GRCh38
rs202165338 Chr.13: 77901232-77901232 on Build GRCh38
rs9530703 Chr.13: 77901329-77901329 on Build GRCh38
rs781583323 Chr.13: 77901145-77901145 on Build GRCh38
rs12720194 Chr.13: 77901280-77901280 on Build GRCh38
rs199937989 Chr.13: 77901189-77901189 on Build GRCh38
rs138785424 Chr.13: 77901299-77901299 on Build GRCh38
rs997676377 Chr.13: 77901242-77901242 on Build GRCh38
rs756306738 Chr.13: 77901246-77901246 on Build GRCh38
rs765690914 Chr.13: 77901215-77901215 on Build GRCh38
rs968336645 Chr.13: 77901331-77901331 on Build GRCh38
rs763342247 Chr.13: 77901222-77901222 on Build GRCh38
rs104894387 Chr.13: 77901181-77901181 on Build GRCh38
rs757944336 Chr.13: 77901180-77901180 on Build GRCh38
rs12720193 Chr.13: 77901346-77901346 on Build GRCh38
rs5351 Chr.13: 77901178-77901178 on Build GRCh38
rs3027105 Chr.13: 77901345-77901345 on Build GRCh38
rs3027103 Chr.13: 77901177-77901177 on Build GRCh38
rs370187673 Chr.13: 77901130-77901130 on Build GRCh38
rs142295388 Chr.13: 77901188-77901188 on Build GRCh38
rs758958773 Chr.13: 77901128-77901128 on Build GRCh38
rs12720195 Chr.13: 77901231-77901231 on Build GRCh38
rs781099239 Chr.13: 77901236-77901236 on Build GRCh38
rs974099624 Chr.13: 77901276-77901276 on Build GRCh38
rs104894389 Chr.13: 77901185-77901185 on Build GRCh38
rs3027104 Chr.13: 77901328-77901328 on Build GRCh38
rs760242225 Chr.13: 77901214-77901214 on Build GRCh38
rs768447144 Chr.13: 77901312-77901312 on Build GRCh38
rs761336304 Chr.13: 77901190-77901190 on Build GRCh38
rs772959328 Chr.13: 77901213-77901213 on Build GRCh38
rs773587906 Chr.13: 77901257-77901264 on Build GRCh38
rs750721433 Chr.13: 77901238-77901238 on Build GRCh38
rs746579742 Chr.13: 77901270-77901270 on Build GRCh38
rs753014932 Chr.13: 77901220-77901220 on Build GRCh38
rs769735757 Chr.13: 77901132-77901132 on Build GRCh38

View Full Product Details