Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101727390-101727639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
UniGene
Hs.444135
Gene Chromosome Location
Chr.3: 101724098-101770562 on Build GRCh38
Transcripts
NM_001303401.1 NM_024548.3 XM_006713743.3 XM_011513127.1 XM_017007178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6095441 c.272G>A p.S91N Chr.3: 101727468-101727468 on Build GRCh38
7173231 c.261G>A p.L87L Chr.3: 101727457-101727457 on Build GRCh38
4759476 c.248G>A p.R83H Chr.3: 101727444-101727444 on Build GRCh38
7261173 c.205C>T p.R69W Chr.3: 101727401-101727401 on Build GRCh38
5509953 c.215G>A p.R72Q Chr.3: 101727411-101727411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755150222 Chr.3: 101727509-101727509 on Build GRCh38
rs770955468 Chr.3: 101727542-101727542 on Build GRCh38
rs762311746 Chr.3: 101727500-101727500 on Build GRCh38
rs145437722 Chr.3: 101727506-101727506 on Build GRCh38
rs536511998 Chr.3: 101727445-101727445 on Build GRCh38
rs960900230 Chr.3: 101727443-101727443 on Build GRCh38
rs200492494 Chr.3: 101727448-101727448 on Build GRCh38
rs765011864 Chr.3: 101727444-101727444 on Build GRCh38
rs371067404 Chr.3: 101727497-101727497 on Build GRCh38
rs762727311 Chr.3: 101727566-101727566 on Build GRCh38
rs776432630 Chr.3: 101727554-101727554 on Build GRCh38
rs184909001 Chr.3: 101727487-101727487 on Build GRCh38
rs748150474 Chr.3: 101727537-101727537 on Build GRCh38
rs143663483 Chr.3: 101727426-101727426 on Build GRCh38
rs373775352 Chr.3: 101727576-101727576 on Build GRCh38
rs374487096 Chr.3: 101727564-101727564 on Build GRCh38
rs780800947 Chr.3: 101727468-101727468 on Build GRCh38
rs750866373 Chr.3: 101727483-101727483 on Build GRCh38
rs774053681 Chr.3: 101727578-101727578 on Build GRCh38
rs761216809 Chr.3: 101727579-101727579 on Build GRCh38
rs745755956 Chr.3: 101727560-101727560 on Build GRCh38
rs770738404 Chr.3: 101727434-101727434 on Build GRCh38
rs1024966245 Chr.3: 101727435-101727435 on Build GRCh38
rs76508572 Chr.3: 101727587-101727587 on Build GRCh38
rs970012345 Chr.3: 101727452-101727452 on Build GRCh38
rs774363921 Chr.3: 101727420-101727420 on Build GRCh38
rs763720418 Chr.3: 101727450-101727450 on Build GRCh38
rs775379206 Chr.3: 101727565-101727565 on Build GRCh38
rs907557036 Chr.3: 101727532-101727532 on Build GRCh38
rs548722311 Chr.3: 101727436-101727436 on Build GRCh38
rs941022620 Chr.3: 101727499-101727499 on Build GRCh38
rs756439484 Chr.3: 101727490-101727490 on Build GRCh38
rs751076429 Chr.3: 101727464-101727464 on Build GRCh38
rs191543705 Chr.3: 101727461-101727461 on Build GRCh38
rs757822875 Chr.3: 101727469-101727469 on Build GRCh38
rs779002101 Chr.3: 101727536-101727536 on Build GRCh38
rs751639054 Chr.3: 101727493-101727493 on Build GRCh38

View Full Product Details