Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114127400-114127651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC186
Entrez Gene ID
55088
Gene Name
coiled-coil domain containing 186
Gene Aliases
C10orf118
UniGene
Hs.159066
Gene Chromosome Location
Chr.10: 114120862-114174312 on Build GRCh38
Transcripts
NM_001321829.1 NM_018017.3 NR_135815.1 XM_011539915.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159442 c.2290A>C p.R764R Chr.10: 114127564-114127564 on Build GRCh38
4011738 c.2332A>C p.K778Q Chr.10: 114127522-114127522 on Build GRCh38
914989 c.2291G>T p.R764I Chr.10: 114127563-114127563 on Build GRCh38
3786323 c.2224G>T p.E742* Chr.10: 114127630-114127630 on Build GRCh38
914990 c.2215C>T p.R739* Chr.10: 114127639-114127639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775775012 Chr.10: 114127612-114127612 on Build GRCh38
rs554420661 Chr.10: 114127616-114127616 on Build GRCh38
rs1034134850 Chr.10: 114127434-114127434 on Build GRCh38
rs777861966 Chr.10: 114127556-114127556 on Build GRCh38
rs768583958 Chr.10: 114127553-114127553 on Build GRCh38
rs146862355 Chr.10: 114127550-114127550 on Build GRCh38
rs746462938 Chr.10: 114127577-114127577 on Build GRCh38
rs756851257 Chr.10: 114127588-114127588 on Build GRCh38
rs750628801 Chr.10: 114127540-114127540 on Build GRCh38
rs369111615 Chr.10: 114127598-114127598 on Build GRCh38
rs201662734 Chr.10: 114127538-114127538 on Build GRCh38
rs776036322 Chr.10: 114127453-114127453 on Build GRCh38
rs553836080 Chr.10: 114127518-114127518 on Build GRCh38
rs199729026 Chr.10: 114127446-114127446 on Build GRCh38
rs200228945 Chr.10: 114127447-114127447 on Build GRCh38
rs767718072 Chr.10: 114127519-114127519 on Build GRCh38
rs992475354 Chr.10: 114127436-114127436 on Build GRCh38
rs140171648 Chr.10: 114127622-114127622 on Build GRCh38
rs564572727 Chr.10: 114127602-114127602 on Build GRCh38
rs780858415 Chr.10: 114127592-114127592 on Build GRCh38
rs959777721 Chr.10: 114127435-114127435 on Build GRCh38
rs375846377 Chr.10: 114127458-114127458 on Build GRCh38
rs746517320 Chr.10: 114127595-114127596 on Build GRCh38
rs774851809 Chr.10: 114127625-114127625 on Build GRCh38
rs751703061 Chr.10: 114127480-114127480 on Build GRCh38
rs763501570 Chr.10: 114127456-114127456 on Build GRCh38
rs752670951 Chr.10: 114127547-114127547 on Build GRCh38
rs79637542 Chr.10: 114127543-114127543 on Build GRCh38
rs140658837 Chr.10: 114127604-114127604 on Build GRCh38

View Full Product Details