Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56055647-56055902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6A
Entrez Gene ID
908
Gene Name
chaperonin containing TCP1 subunit 6A
Gene Aliases
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20
UniGene
Hs.82916
Gene Chromosome Location
Chr.7: 56051685-56063989 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279698 c.464C>A p.S155Y Chr.7: 56055751-56055751 on Build GRCh38
5635257 c.494C>T p.A165V Chr.7: 56055781-56055781 on Build GRCh38
3881898 c.471T>C p.R157R Chr.7: 56055758-56055758 on Build GRCh38
7153860 c.371A>T p.E124V Chr.7: 56055658-56055658 on Build GRCh38
5492234 c.470G>A p.R157H Chr.7: 56055757-56055757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764557858 Chr.7: 56055844-56055846 on Build GRCh38
rs751493247 Chr.7: 56055783-56055783 on Build GRCh38
rs779079031 Chr.7: 56055825-56055825 on Build GRCh38
rs754783600 Chr.7: 56055693-56055693 on Build GRCh38
rs914022025 Chr.7: 56055820-56055820 on Build GRCh38
rs746209990 Chr.7: 56055751-56055751 on Build GRCh38
rs73343772 Chr.7: 56055816-56055816 on Build GRCh38
rs368279676 Chr.7: 56055830-56055830 on Build GRCh38
rs562140324 Chr.7: 56055874-56055874 on Build GRCh38
rs761395472 Chr.7: 56055757-56055757 on Build GRCh38
rs772701047 Chr.7: 56055770-56055770 on Build GRCh38
rs756428482 Chr.7: 56055713-56055713 on Build GRCh38
rs368585749 Chr.7: 56055788-56055788 on Build GRCh38
rs756338375 Chr.7: 56055809-56055809 on Build GRCh38
rs767372513 Chr.7: 56055801-56055801 on Build GRCh38
rs181411778 Chr.7: 56055838-56055838 on Build GRCh38
rs757320999 Chr.7: 56055822-56055822 on Build GRCh38
rs755910615 Chr.7: 56055818-56055818 on Build GRCh38
rs749384523 Chr.7: 56055731-56055731 on Build GRCh38
rs769437110 Chr.7: 56055768-56055768 on Build GRCh38
rs762431193 Chr.7: 56055780-56055780 on Build GRCh38
rs200144880 Chr.7: 56055756-56055756 on Build GRCh38
rs371873192 Chr.7: 56055806-56055806 on Build GRCh38
rs759552800 Chr.7: 56055784-56055784 on Build GRCh38
rs201108756 Chr.7: 56055781-56055781 on Build GRCh38
rs763343329 Chr.7: 56055807-56055807 on Build GRCh38
rs544013814 Chr.7: 56055867-56055867 on Build GRCh38
rs529251204 Chr.7: 56055811-56055811 on Build GRCh38
rs779338195 Chr.7: 56055736-56055736 on Build GRCh38
rs1043790684 Chr.7: 56055861-56055861 on Build GRCh38
rs772283442 Chr.7: 56055753-56055753 on Build GRCh38
rs969718475 Chr.7: 56055749-56055749 on Build GRCh38
rs769419043 Chr.7: 56055849-56055849 on Build GRCh38
rs752556289 Chr.7: 56055804-56055804 on Build GRCh38
rs752576116 Chr.7: 56055796-56055796 on Build GRCh38
rs772643463 Chr.7: 56055831-56055831 on Build GRCh38
rs553391026 Chr.7: 56055718-56055718 on Build GRCh38
rs945439678 Chr.7: 56055855-56055855 on Build GRCh38
rs751359684 Chr.7: 56055677-56055677 on Build GRCh38
rs780327719 Chr.7: 56055847-56055847 on Build GRCh38
rs145709363 Chr.7: 56055703-56055703 on Build GRCh38
rs201362156 Chr.7: 56055733-56055733 on Build GRCh38
rs781536224 Chr.7: 56055695-56055695 on Build GRCh38
rs747436268 Chr.7: 56055848-56055848 on Build GRCh38
rs879176516 Chr.7: 56055872-56055872 on Build GRCh38

View Full Product Details