Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 664504-664777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH1
Entrez Gene ID
157697
Gene Name
glutamate rich 1
Gene Aliases
HSPC319
UniGene
Hs.655310
Gene Chromosome Location
Chr.8: 614737-731239 on Build GRCh38
Transcripts
NM_001303100.1 NM_207332.2 XM_006716234.3 XM_011534731.2 XM_011534735.2 XM_011534739.2 XM_017013114.1 XM_017013115.1 XM_017013116.1 XM_017013117.1 XM_017013118.1 XM_017013119.1 XM_017013120.1 XM_017013121.1 XM_017013122.1 XM_017013123.1 XM_017013124.1 XM_017013125.1 XM_017013126.1 XM_017013127.1 XM_017013132.1 XM_017013134.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7264395 c.1259-3T>A p.? Chr.8: 664679-664679 on Build GRCh38
3900846 c.1307T>A p.I436N Chr.8: 664628-664628 on Build GRCh38
751086 c.1303C>A p.H435N Chr.8: 664632-664632 on Build GRCh38
6295229 c.1261C>A p.H421N Chr.8: 664674-664674 on Build GRCh38
1457668 c.1327G>A p.D443N Chr.8: 664608-664608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186841311 Chr.8: 664592-664592 on Build GRCh38
rs374813242 Chr.8: 664604-664604 on Build GRCh38
rs942160211 Chr.8: 664675-664675 on Build GRCh38
rs561913656 Chr.8: 664537-664537 on Build GRCh38
rs752885695 Chr.8: 664608-664608 on Build GRCh38
rs771298745 Chr.8: 664672-664672 on Build GRCh38
rs781679491 Chr.8: 664578-664578 on Build GRCh38
rs756100695 Chr.8: 664650-664650 on Build GRCh38
rs751761333 Chr.8: 664620-664620 on Build GRCh38
rs908011963 Chr.8: 664656-664656 on Build GRCh38
rs763087324 Chr.8: 664690-664690 on Build GRCh38
rs753865663 Chr.8: 664631-664632 on Build GRCh38
rs1037743191 Chr.8: 664707-664707 on Build GRCh38
rs917441806 Chr.8: 664568-664568 on Build GRCh38
rs540763117 Chr.8: 664732-664732 on Build GRCh38
rs559567912 Chr.8: 664753-664753 on Build GRCh38
rs200656329 Chr.8: 664575-664575 on Build GRCh38
rs754961249 Chr.8: 664695-664695 on Build GRCh38
rs773874964 Chr.8: 664587-664587 on Build GRCh38
rs748924815 Chr.8: 664574-664574 on Build GRCh38
rs781305846 Chr.8: 664628-664628 on Build GRCh38
rs760003353 Chr.8: 664593-664593 on Build GRCh38
rs748726274 Chr.8: 664653-664653 on Build GRCh38
rs763142371 Chr.8: 664610-664610 on Build GRCh38
rs759579583 Chr.8: 664739-664739 on Build GRCh38
rs780900867 Chr.8: 664626-664626 on Build GRCh38
rs762005917 Chr.8: 664722-664722 on Build GRCh38
rs747802456 Chr.8: 664664-664664 on Build GRCh38
rs890327528 Chr.8: 664737-664737 on Build GRCh38
rs776145757 Chr.8: 664688-664688 on Build GRCh38
rs113707327 Chr.8: 664660-664660 on Build GRCh38
rs770371327 Chr.8: 664685-664685 on Build GRCh38
rs769261366 Chr.8: 664555-664555 on Build GRCh38
rs570997850 Chr.8: 664560-664560 on Build GRCh38
rs750367517 Chr.8: 664636-664636 on Build GRCh38
rs201294062 Chr.8: 664588-664588 on Build GRCh38
rs989895248 Chr.8: 664531-664531 on Build GRCh38
rs751785425 Chr.8: 664706-664706 on Build GRCh38
rs200071096 Chr.8: 664695-664695 on Build GRCh38
rs563145724 Chr.8: 664625-664625 on Build GRCh38
rs764345933 Chr.8: 664613-664613 on Build GRCh38
rs779979642 Chr.8: 664651-664651 on Build GRCh38
rs200450936 Chr.8: 664606-664606 on Build GRCh38
rs898924006 Chr.8: 664754-664754 on Build GRCh38
rs368038515 Chr.8: 664654-664654 on Build GRCh38
rs760110981 Chr.8: 664684-664684 on Build GRCh38
rs1051668629 Chr.8: 664731-664731 on Build GRCh38
rs775046150 Chr.8: 664559-664559 on Build GRCh38
rs765103730 Chr.8: 664745-664745 on Build GRCh38
rs778907655 Chr.8: 664720-664721 on Build GRCh38
rs769712076 Chr.8: 664673-664673 on Build GRCh38

View Full Product Details