Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2251453-2251686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECI1
Entrez Gene ID
1632
Gene Name
enoyl-CoA delta isomerase 1
Gene Aliases
DCI
UniGene
Hs.403436
Gene Chromosome Location
Chr.16: 2239872-2251601 on Build GRCh38
Transcripts
NM_001178029.1 NM_001919.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4550365 c.49G>A p.A17T Chr.16: 2251518-2251518 on Build GRCh38
5627478 c.40C>G p.L14V Chr.16: 2251527-2251527 on Build GRCh38
434911 c.48C>G p.R16R Chr.16: 2251519-2251519 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199944223 Chr.16: 2251526-2251526 on Build GRCh38
rs772468885 Chr.16: 2251483-2251483 on Build GRCh38
rs772827843 Chr.16: 2251515-2251515 on Build GRCh38
rs900211278 Chr.16: 2251560-2251560 on Build GRCh38
rs530086009 Chr.16: 2251620-2251620 on Build GRCh38
rs984926166 Chr.16: 2251566-2251566 on Build GRCh38
rs754862311 Chr.16: 2251523-2251523 on Build GRCh38
rs746202495 Chr.16: 2251578-2251578 on Build GRCh38
rs774121673 Chr.16: 2251612-2251612 on Build GRCh38
rs550289429 Chr.16: 2251630-2251630 on Build GRCh38
rs561290323 Chr.16: 2251604-2251604 on Build GRCh38
rs904419465 Chr.16: 2251517-2251517 on Build GRCh38
rs774613604 Chr.16: 2251475-2251475 on Build GRCh38
rs959113454 Chr.16: 2251554-2251554 on Build GRCh38
rs199982693 Chr.16: 2251598-2251598 on Build GRCh38
rs948822255 Chr.16: 2251668-2251668 on Build GRCh38
rs778024807 Chr.16: 2251538-2251538 on Build GRCh38
rs572664579 Chr.16: 2251591-2251591 on Build GRCh38
rs868120216 Chr.16: 2251582-2251582 on Build GRCh38
rs777908966 Chr.16: 2251516-2251516 on Build GRCh38
rs1015730243 Chr.16: 2251630-2251632 on Build GRCh38
rs988881339 Chr.16: 2251504-2251504 on Build GRCh38
rs981630859 Chr.16: 2251522-2251522 on Build GRCh38
rs772551198 Chr.16: 2251584-2251584 on Build GRCh38
rs1006665698 Chr.16: 2251474-2251474 on Build GRCh38
rs1007130560 Chr.16: 2251662-2251662 on Build GRCh38
rs749484668 Chr.16: 2251545-2251545 on Build GRCh38
rs113641186 Chr.16: 2251511-2251511 on Build GRCh38
rs757560123 Chr.16: 2251573-2251573 on Build GRCh38
rs915982514 Chr.16: 2251657-2251657 on Build GRCh38
rs182577651 Chr.16: 2251585-2251585 on Build GRCh38
rs1004796054 Chr.16: 2251629-2251630 on Build GRCh38
rs779276270 Chr.16: 2251574-2251574 on Build GRCh38
rs746322253 Chr.16: 2251477-2251477 on Build GRCh38
rs761303776 Chr.16: 2251493-2251493 on Build GRCh38
rs756244379 Chr.16: 2251527-2251527 on Build GRCh38
rs776120071 Chr.16: 2251487-2251487 on Build GRCh38
rs933912653 Chr.16: 2251622-2251622 on Build GRCh38
rs186632369 Chr.16: 2251660-2251660 on Build GRCh38
rs563807767 Chr.16: 2251653-2251653 on Build GRCh38
rs992945500 Chr.16: 2251638-2251638 on Build GRCh38
rs11541763 Chr.16: 2251519-2251519 on Build GRCh38
rs1043393759 Chr.16: 2251500-2251500 on Build GRCh38
rs765936249 Chr.16: 2251607-2251607 on Build GRCh38
rs889653743 Chr.16: 2251652-2251652 on Build GRCh38
rs576283920 Chr.16: 2251669-2251669 on Build GRCh38

View Full Product Details