Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110015340-110015613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCYL1
Entrez Gene ID
10768
Gene Name
adenosylhomocysteinase like 1
Gene Aliases
DCAL, IRBIT, PPP1R78, PRO0233, XPVKONA
UniGene
Hs.743973
Gene Chromosome Location
Chr.1: 109984741-110023742 on Build GRCh38
Transcripts
NM_001242673.1 NM_001242674.1 NM_001242675.1 NM_001242676.1 NM_006621.5 XM_011540535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457587 c.679C>T p.L227L Chr.1: 110015428-110015428 on Build GRCh38
6842937 c.774T>A p.G258G Chr.1: 110015523-110015523 on Build GRCh38
4020453 c.749G>A p.G250D Chr.1: 110015498-110015498 on Build GRCh38
1747607 c.744C>T p.I248I Chr.1: 110015493-110015493 on Build GRCh38
893598 c.694G>T p.D232Y Chr.1: 110015443-110015443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747463826 Chr.1: 110015495-110015495 on Build GRCh38
rs191023484 Chr.1: 110015588-110015588 on Build GRCh38
rs373997078 Chr.1: 110015395-110015395 on Build GRCh38
rs752938069 Chr.1: 110015377-110015377 on Build GRCh38
rs749113861 Chr.1: 110015370-110015370 on Build GRCh38
rs534866658 Chr.1: 110015587-110015587 on Build GRCh38
rs367786831 Chr.1: 110015479-110015479 on Build GRCh38
rs765440952 Chr.1: 110015376-110015376 on Build GRCh38
rs1040996894 Chr.1: 110015385-110015385 on Build GRCh38
rs777015533 Chr.1: 110015537-110015537 on Build GRCh38
rs199628026 Chr.1: 110015421-110015421 on Build GRCh38
rs780401097 Chr.1: 110015583-110015583 on Build GRCh38
rs999088373 Chr.1: 110015373-110015373 on Build GRCh38
rs755606831 Chr.1: 110015478-110015478 on Build GRCh38
rs895316925 Chr.1: 110015384-110015384 on Build GRCh38
rs757286940 Chr.1: 110015416-110015416 on Build GRCh38
rs1034299294 Chr.1: 110015371-110015371 on Build GRCh38
rs772538545 Chr.1: 110015494-110015494 on Build GRCh38
rs150954522 Chr.1: 110015451-110015451 on Build GRCh38
rs759612161 Chr.1: 110015560-110015560 on Build GRCh38
rs370154404 Chr.1: 110015502-110015502 on Build GRCh38
rs1005296459 Chr.1: 110015585-110015585 on Build GRCh38
rs748985029 Chr.1: 110015493-110015493 on Build GRCh38
rs762705532 Chr.1: 110015382-110015382 on Build GRCh38
rs751487720 Chr.1: 110015407-110015407 on Build GRCh38
rs769794983 Chr.1: 110015566-110015566 on Build GRCh38
rs185084590 Chr.1: 110015567-110015567 on Build GRCh38
rs996487291 Chr.1: 110015559-110015559 on Build GRCh38
rs1003724194 Chr.1: 110015369-110015369 on Build GRCh38

View Full Product Details