Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2539777-2540006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAT1
Entrez Gene ID
221927
Gene Name
BRCA1 associated ATM activator 1
Gene Aliases
BAAT1, C7orf27, RMFSL
UniGene
Hs.520623
Gene Chromosome Location
Chr.7: 2537810-2555758 on Build GRCh38
Transcripts
NM_152743.3 XM_005249643.4 XM_011515177.2 XM_011515178.1 XM_011515179.2 XM_011515181.2 XM_011515182.2 XM_011515183.1 XM_011515184.2 XM_011515186.2 XM_017011833.1 XM_017011834.1 XM_017011835.1 XM_017011836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922922 c.1439G>A p.S480N Chr.7: 2539845-2539845 on Build GRCh38
5997334 c.1480A>G p.I494V Chr.7: 2539804-2539804 on Build GRCh38
5451903 c.1396-7G>A p.? Chr.7: 2539895-2539895 on Build GRCh38
6377137 c.1471G>A p.G491S Chr.7: 2539813-2539813 on Build GRCh38
6285879 c.1498+6G>T p.? Chr.7: 2539780-2539780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755683091 Chr.7: 2539935-2539935 on Build GRCh38
rs374233073 Chr.7: 2539838-2539838 on Build GRCh38
rs371170298 Chr.7: 2539874-2539874 on Build GRCh38
rs767244180 Chr.7: 2539926-2539926 on Build GRCh38
rs770888883 Chr.7: 2539938-2539941 on Build GRCh38
rs749881547 Chr.7: 2539837-2539837 on Build GRCh38
rs1047734135 Chr.7: 2539904-2539904 on Build GRCh38
rs529330648 Chr.7: 2539987-2539987 on Build GRCh38
rs769634441 Chr.7: 2539956-2539956 on Build GRCh38
rs748374616 Chr.7: 2539941-2539942 on Build GRCh38
rs148923187 Chr.7: 2539863-2539863 on Build GRCh38
rs906327946 Chr.7: 2539823-2539823 on Build GRCh38
rs560491630 Chr.7: 2539976-2539976 on Build GRCh38
rs988671301 Chr.7: 2539959-2539959 on Build GRCh38
rs762040027 Chr.7: 2539812-2539812 on Build GRCh38
rs577945739 Chr.7: 2539800-2539800 on Build GRCh38
rs952381868 Chr.7: 2539850-2539850 on Build GRCh38
rs896741988 Chr.7: 2539911-2539911 on Build GRCh38
rs913159579 Chr.7: 2539824-2539824 on Build GRCh38
rs546991713 Chr.7: 2539947-2539947 on Build GRCh38
rs945491251 Chr.7: 2539975-2539975 on Build GRCh38
rs565153096 Chr.7: 2539920-2539920 on Build GRCh38
rs906407872 Chr.7: 2539985-2539985 on Build GRCh38
rs200500603 Chr.7: 2539899-2539899 on Build GRCh38
rs986767030 Chr.7: 2539840-2539840 on Build GRCh38
rs576174160 Chr.7: 2539895-2539895 on Build GRCh38
rs533528841 Chr.7: 2539861-2539861 on Build GRCh38
rs772904982 Chr.7: 2539852-2539852 on Build GRCh38
rs775626426 Chr.7: 2539884-2539884 on Build GRCh38
rs749698079 Chr.7: 2539799-2539799 on Build GRCh38
rs145727035 Chr.7: 2539817-2539817 on Build GRCh38
rs752652217 Chr.7: 2539821-2539821 on Build GRCh38
rs555986902 Chr.7: 2539829-2539829 on Build GRCh38
rs761392479 Chr.7: 2539921-2539921 on Build GRCh38
rs1020119858 Chr.7: 2539971-2539971 on Build GRCh38
rs1005210342 Chr.7: 2539859-2539859 on Build GRCh38
rs771129655 Chr.7: 2539880-2539880 on Build GRCh38
rs773303692 Chr.7: 2539813-2539813 on Build GRCh38
rs780893968 Chr.7: 2539848-2539848 on Build GRCh38
rs200248064 Chr.7: 2539833-2539833 on Build GRCh38
rs553366638 Chr.7: 2539862-2539862 on Build GRCh38
rs780596356 Chr.7: 2539836-2539836 on Build GRCh38
rs199717809 Chr.7: 2539927-2539927 on Build GRCh38
rs539902160 Chr.7: 2539804-2539804 on Build GRCh38
rs150268284 Chr.7: 2539806-2539806 on Build GRCh38
rs562500791 Chr.7: 2539879-2539879 on Build GRCh38
rs542709547 Chr.7: 2539878-2539878 on Build GRCh38
rs936893514 Chr.7: 2539902-2539902 on Build GRCh38
rs763936681 Chr.7: 2539828-2539828 on Build GRCh38
rs762907691 Chr.7: 2539826-2539826 on Build GRCh38

View Full Product Details