Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48014100-48014373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELSPBP1
Entrez Gene ID
64100
Gene Name
epididymal sperm binding protein 1
Gene Aliases
E12, EDDM12, EL149, HE12
UniGene
Hs.104894
Gene Chromosome Location
Chr.19: 47994651-48025153 on Build GRCh38
Transcripts
NM_022142.4 XM_017027130.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1394977 c.208G>T p.D70Y Chr.19: 48014308-48014308 on Build GRCh38
3536405 c.82G>A p.E28K Chr.19: 48014182-48014182 on Build GRCh38
7113773 c.181G>A p.G61S Chr.19: 48014281-48014281 on Build GRCh38
567205 c.110G>A p.G37E Chr.19: 48014210-48014210 on Build GRCh38
3422959 c.95C>A p.P32H Chr.19: 48014195-48014195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042338655 Chr.19: 48014285-48014285 on Build GRCh38
rs757186437 Chr.19: 48014163-48014163 on Build GRCh38
rs147073337 Chr.19: 48014271-48014271 on Build GRCh38
rs753173798 Chr.19: 48014251-48014251 on Build GRCh38
rs750324752 Chr.19: 48014274-48014274 on Build GRCh38
rs781628866 Chr.19: 48014194-48014194 on Build GRCh38
rs778253442 Chr.19: 48014211-48014211 on Build GRCh38
rs775178888 Chr.19: 48014241-48014241 on Build GRCh38
rs973904946 Chr.19: 48014244-48014244 on Build GRCh38
rs751607695 Chr.19: 48014175-48014175 on Build GRCh38
rs765790649 Chr.19: 48014328-48014328 on Build GRCh38
rs779504982 Chr.19: 48014319-48014319 on Build GRCh38
rs745306365 Chr.19: 48014233-48014233 on Build GRCh38
rs748367789 Chr.19: 48014202-48014202 on Build GRCh38
rs369872763 Chr.19: 48014281-48014282 on Build GRCh38
rs989303155 Chr.19: 48014277-48014277 on Build GRCh38
rs77097016 Chr.19: 48014279-48014279 on Build GRCh38
rs768247053 Chr.19: 48014330-48014330 on Build GRCh38
rs951806429 Chr.19: 48014160-48014160 on Build GRCh38
rs62129078 Chr.19: 48014281-48014281 on Build GRCh38
rs755167342 Chr.19: 48014185-48014185 on Build GRCh38
rs771106886 Chr.19: 48014185-48014186 on Build GRCh38
rs371933085 Chr.19: 48014132-48014132 on Build GRCh38
rs188923874 Chr.19: 48014308-48014308 on Build GRCh38
rs537927439 Chr.19: 48014318-48014318 on Build GRCh38
rs370179993 Chr.19: 48014334-48014334 on Build GRCh38
rs866242476 Chr.19: 48014249-48014249 on Build GRCh38
rs756597821 Chr.19: 48014207-48014207 on Build GRCh38
rs775445007 Chr.19: 48014140-48014140 on Build GRCh38
rs201798268 Chr.19: 48014214-48014214 on Build GRCh38
rs117586587 Chr.19: 48014238-48014238 on Build GRCh38
rs766398774 Chr.19: 48014171-48014171 on Build GRCh38
rs771423768 Chr.19: 48014222-48014222 on Build GRCh38
rs200336076 Chr.19: 48014309-48014309 on Build GRCh38
rs778306785 Chr.19: 48014291-48014291 on Build GRCh38
rs138110467 Chr.19: 48014280-48014280 on Build GRCh38
rs150199608 Chr.19: 48014184-48014184 on Build GRCh38
rs541922409 Chr.19: 48014129-48014129 on Build GRCh38
rs768951502 Chr.19: 48014181-48014181 on Build GRCh38
rs908305917 Chr.19: 48014226-48014226 on Build GRCh38
rs145096514 Chr.19: 48014272-48014272 on Build GRCh38
rs760133348 Chr.19: 48014299-48014300 on Build GRCh38
rs959690084 Chr.19: 48014142-48014142 on Build GRCh38
rs141635603 Chr.19: 48014265-48014265 on Build GRCh38
rs376690556 Chr.19: 48014324-48014324 on Build GRCh38
rs773750317 Chr.19: 48014231-48014231 on Build GRCh38
rs776784350 Chr.19: 48014254-48014256 on Build GRCh38
rs754176515 Chr.19: 48014298-48014298 on Build GRCh38
rs41494755 Chr.19: 48014130-48014130 on Build GRCh38
rs375402318 Chr.19: 48014165-48014165 on Build GRCh38

View Full Product Details