Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 182729835-182730076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
UniGene
Hs.516632
Gene Chromosome Location
Chr.2: 182716041-182780025 on Build GRCh38
Transcripts
NM_001271581.1 NM_018981.2 NR_073365.1 NR_073366.1 NR_073367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1691663 c.701G>A p.R234K Chr.2: 182729915-182729915 on Build GRCh38
1631645 c.650A>G p.H217R Chr.2: 182729864-182729864 on Build GRCh38
4413696 c.715G>C p.E239Q Chr.2: 182729929-182729929 on Build GRCh38
3990905 c.678G>T p.V226V Chr.2: 182729892-182729892 on Build GRCh38
7086303 c.652G>A p.G218R Chr.2: 182729866-182729866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568147630 Chr.2: 182730026-182730026 on Build GRCh38
rs551127442 Chr.2: 182729897-182729897 on Build GRCh38
rs61748252 Chr.2: 182729894-182729894 on Build GRCh38
rs141906022 Chr.2: 182729920-182729920 on Build GRCh38
rs552700591 Chr.2: 182730012-182730012 on Build GRCh38
rs771758358 Chr.2: 182729861-182729863 on Build GRCh38
rs762726984 Chr.2: 182729873-182729873 on Build GRCh38
rs962653142 Chr.2: 182730029-182730029 on Build GRCh38
rs1041708444 Chr.2: 182730016-182730016 on Build GRCh38
rs185305206 Chr.2: 182729972-182729972 on Build GRCh38
rs191829754 Chr.2: 182729939-182729939 on Build GRCh38
rs369375869 Chr.2: 182729902-182729902 on Build GRCh38
rs767002657 Chr.2: 182729922-182729923 on Build GRCh38
rs182412137 Chr.2: 182729950-182729950 on Build GRCh38
rs955736016 Chr.2: 182729921-182729921 on Build GRCh38
rs750240525 Chr.2: 182729865-182729865 on Build GRCh38
rs578205322 Chr.2: 182730044-182730044 on Build GRCh38
rs753174442 Chr.2: 182729975-182729975 on Build GRCh38
rs914786525 Chr.2: 182729940-182729940 on Build GRCh38
rs987416716 Chr.2: 182729885-182729885 on Build GRCh38
rs372625469 Chr.2: 182729909-182729909 on Build GRCh38
rs1002940143 Chr.2: 182730031-182730031 on Build GRCh38
rs974430261 Chr.2: 182730047-182730047 on Build GRCh38
rs536194538 Chr.2: 182730011-182730011 on Build GRCh38
rs538757633 Chr.2: 182730036-182730036 on Build GRCh38
rs376902618 Chr.2: 182729961-182729961 on Build GRCh38
rs140936889 Chr.2: 182729857-182729857 on Build GRCh38
rs1040286255 Chr.2: 182729876-182729876 on Build GRCh38
rs757794320 Chr.2: 182729910-182729910 on Build GRCh38
rs374998022 Chr.2: 182729863-182729863 on Build GRCh38
rs780370802 Chr.2: 182729942-182729942 on Build GRCh38
rs776707863 Chr.2: 182729964-182729964 on Build GRCh38
rs766264424 Chr.2: 182729878-182729878 on Build GRCh38
rs145832696 Chr.2: 182730042-182730042 on Build GRCh38

View Full Product Details