Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184382562-184382821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRD
Entrez Gene ID
8646
Gene Name
chordin
Gene Aliases
-
UniGene
Hs.166186
Gene Chromosome Location
Chr.3: 184380073-184389835 on Build GRCh38
Transcripts
NM_001304472.1 NM_001304473.1 NM_001304474.1 NM_003741.3 NR_130747.1 XM_011513254.1 XM_011513256.1 XM_017007388.1 XM_017007389.1 XM_017007390.1 XM_017007391.1 XM_017007392.1 XM_017007393.1 XM_017007394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1284123 c.855C>T p.A285A Chr.3: 184382647-184382647 on Build GRCh38
6031502 c.939delT p.L315fs*15 Chr.3: 184382731-184382731 on Build GRCh38
1693892 c.877C>T p.P293S Chr.3: 184382669-184382669 on Build GRCh38
5344425 c.897C>A p.G299G Chr.3: 184382689-184382689 on Build GRCh38
730204 c.904C>A p.L302M Chr.3: 184382696-184382696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756777032 Chr.3: 184382592-184382592 on Build GRCh38
rs905459510 Chr.3: 184382730-184382730 on Build GRCh38
rs139538414 Chr.3: 184382703-184382703 on Build GRCh38
rs191770213 Chr.3: 184382725-184382725 on Build GRCh38
rs376828858 Chr.3: 184382689-184382689 on Build GRCh38
rs762183304 Chr.3: 184382748-184382748 on Build GRCh38
rs771133956 Chr.3: 184382629-184382629 on Build GRCh38
rs765865447 Chr.3: 184382774-184382774 on Build GRCh38
rs779699590 Chr.3: 184382612-184382612 on Build GRCh38
rs768926833 Chr.3: 184382742-184382742 on Build GRCh38
rs73887470 Chr.3: 184382798-184382798 on Build GRCh38
rs761071202 Chr.3: 184382652-184382652 on Build GRCh38
rs776802461 Chr.3: 184382747-184382747 on Build GRCh38
rs547793649 Chr.3: 184382642-184382642 on Build GRCh38
rs971413716 Chr.3: 184382791-184382791 on Build GRCh38
rs368929634 Chr.3: 184382705-184382705 on Build GRCh38
rs748096210 Chr.3: 184382626-184382626 on Build GRCh38
rs754937500 Chr.3: 184382587-184382588 on Build GRCh38
rs200378509 Chr.3: 184382590-184382590 on Build GRCh38
rs375530942 Chr.3: 184382681-184382681 on Build GRCh38
rs938225393 Chr.3: 184382732-184382732 on Build GRCh38
rs764422897 Chr.3: 184382658-184382658 on Build GRCh38
rs754791096 Chr.3: 184382700-184382700 on Build GRCh38
rs142141601 Chr.3: 184382728-184382728 on Build GRCh38
rs745959400 Chr.3: 184382637-184382637 on Build GRCh38
rs74825990 Chr.3: 184382799-184382799 on Build GRCh38
rs12629907 Chr.3: 184382602-184382602 on Build GRCh38
rs368180932 Chr.3: 184382607-184382607 on Build GRCh38
rs754350381 Chr.3: 184382591-184382591 on Build GRCh38
rs376490944 Chr.3: 184382606-184382606 on Build GRCh38
rs765982871 Chr.3: 184382698-184382698 on Build GRCh38
rs901103484 Chr.3: 184382663-184382663 on Build GRCh38
rs549778814 Chr.3: 184382588-184382588 on Build GRCh38
rs775607714 Chr.3: 184382648-184382648 on Build GRCh38
rs532416342 Chr.3: 184382764-184382764 on Build GRCh38
rs370155085 Chr.3: 184382762-184382762 on Build GRCh38
rs747261211 Chr.3: 184382741-184382741 on Build GRCh38
rs780919320 Chr.3: 184382701-184382701 on Build GRCh38
rs368023006 Chr.3: 184382771-184382771 on Build GRCh38
rs768373249 Chr.3: 184382614-184382614 on Build GRCh38
rs201596243 Chr.3: 184382620-184382620 on Build GRCh38
rs975262716 Chr.3: 184382675-184382675 on Build GRCh38
rs374822120 Chr.3: 184382753-184382753 on Build GRCh38
rs778460731 Chr.3: 184382597-184382597 on Build GRCh38
rs77289593 Chr.3: 184382695-184382695 on Build GRCh38
rs914165465 Chr.3: 184382588-184382588 on Build GRCh38
rs774509862 Chr.3: 184382630-184382630 on Build GRCh38
rs1020988459 Chr.3: 184382593-184382593 on Build GRCh38
rs931125688 Chr.3: 184382686-184382686 on Build GRCh38
rs373031272 Chr.3: 184382715-184382715 on Build GRCh38
rs201668530 Chr.3: 184382589-184382589 on Build GRCh38
rs751222864 Chr.3: 184382781-184382781 on Build GRCh38
rs745903696 Chr.3: 184382719-184382719 on Build GRCh38
rs747667508 Chr.3: 184382661-184382670 on Build GRCh38
rs780609983 Chr.3: 184382659-184382659 on Build GRCh38
rs777189939 Chr.3: 184382680-184382680 on Build GRCh38
rs868629934 Chr.3: 184382771-184382771 on Build GRCh38

View Full Product Details