Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143300031-143300282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP2
Entrez Gene ID
835
Gene Name
caspase 2
Gene Aliases
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57
UniGene
Hs.368982
Gene Chromosome Location
Chr.7: 143288215-143307696 on Build GRCh38
Transcripts
NM_001224.4 NM_032982.3 NM_032983.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153924 c.871C>T p.L291F Chr.7: 143300046-143300046 on Build GRCh38
6854882 c.924A>G p.L308L Chr.7: 143300251-143300251 on Build GRCh38
6620573 c.907G>A p.A303T Chr.7: 143300234-143300234 on Build GRCh38
6252623 c.877-1G>T p.? Chr.7: 143300203-143300203 on Build GRCh38
5712435 c.916C>T p.P306S Chr.7: 143300243-143300243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752018469 Chr.7: 143300151-143300151 on Build GRCh38
rs763497851 Chr.7: 143300206-143300206 on Build GRCh38
rs886860384 Chr.7: 143300246-143300246 on Build GRCh38
rs370387103 Chr.7: 143300056-143300056 on Build GRCh38
rs200353443 Chr.7: 143300238-143300238 on Build GRCh38
rs145310616 Chr.7: 143300060-143300060 on Build GRCh38
rs767747070 Chr.7: 143300230-143300230 on Build GRCh38
rs746698551 Chr.7: 143300173-143300173 on Build GRCh38
rs376462450 Chr.7: 143300130-143300130 on Build GRCh38
rs762173952 Chr.7: 143300223-143300223 on Build GRCh38
rs756201685 Chr.7: 143300234-143300234 on Build GRCh38
rs937277837 Chr.7: 143300090-143300090 on Build GRCh38
rs754519632 Chr.7: 143300251-143300251 on Build GRCh38
rs751589935 Chr.7: 143300219-143300220 on Build GRCh38
rs765386737 Chr.7: 143300169-143300178 on Build GRCh38
rs960182436 Chr.7: 143300134-143300134 on Build GRCh38
rs752465451 Chr.7: 143300112-143300112 on Build GRCh38
rs745356050 Chr.7: 143300186-143300186 on Build GRCh38
rs776129080 Chr.7: 143300200-143300200 on Build GRCh38
rs200084042 Chr.7: 143300072-143300072 on Build GRCh38
rs774806131 Chr.7: 143300213-143300213 on Build GRCh38
rs754910726 Chr.7: 143300076-143300076 on Build GRCh38
rs764560560 Chr.7: 143300212-143300212 on Build GRCh38
rs770571274 Chr.7: 143300174-143300174 on Build GRCh38
rs747990841 Chr.7: 143300091-143300091 on Build GRCh38
rs766307129 Chr.7: 143300237-143300237 on Build GRCh38
rs776057327 Chr.7: 143300177-143300177 on Build GRCh38
rs759592587 Chr.7: 143300102-143300103 on Build GRCh38
rs750534880 Chr.7: 143300233-143300233 on Build GRCh38
rs372542250 Chr.7: 143300199-143300199 on Build GRCh38
rs368359334 Chr.7: 143300160-143300160 on Build GRCh38
rs761799919 Chr.7: 143300224-143300225 on Build GRCh38
rs777476131 Chr.7: 143300161-143300161 on Build GRCh38
rs778901219 Chr.7: 143300077-143300077 on Build GRCh38
rs374230679 Chr.7: 143300239-143300239 on Build GRCh38

View Full Product Details