Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62390794-62391067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABLES2
Entrez Gene ID
81928
Gene Name
Cdk5 and Abl enzyme substrate 2
Gene Aliases
C20orf150, dJ908M14.2, ik3-2
UniGene
Hs.301040
Gene Chromosome Location
Chr.20: 62388630-62407283 on Build GRCh38
Transcripts
NM_031215.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297233 c.1429C>A p.Q477K Chr.20: 62390979-62390979 on Build GRCh38
7321742 c.1372C>T p.L458L Chr.20: 62391036-62391036 on Build GRCh38
6617210 c.1359C>T p.L453L Chr.20: 62391049-62391049 on Build GRCh38
1028887 c.1418G>A p.R473H Chr.20: 62390990-62390990 on Build GRCh38
229841 c.1408C>A p.H470N Chr.20: 62391000-62391000 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182894361 Chr.20: 62391043-62391043 on Build GRCh38
rs376321988 Chr.20: 62390936-62390936 on Build GRCh38
rs771646874 Chr.20: 62391034-62391034 on Build GRCh38
rs763201175 Chr.20: 62390961-62390961 on Build GRCh38
rs764277990 Chr.20: 62391003-62391004 on Build GRCh38
rs576077601 Chr.20: 62390990-62390990 on Build GRCh38
rs1052493660 Chr.20: 62390850-62390850 on Build GRCh38
rs759173160 Chr.20: 62390922-62390922 on Build GRCh38
rs997964200 Chr.20: 62390901-62390901 on Build GRCh38
rs934306489 Chr.20: 62390834-62390834 on Build GRCh38
rs774897866 Chr.20: 62391036-62391036 on Build GRCh38
rs752396358 Chr.20: 62390869-62390869 on Build GRCh38
rs1040988642 Chr.20: 62390898-62390898 on Build GRCh38
rs1046641561 Chr.20: 62390842-62390842 on Build GRCh38
rs369669045 Chr.20: 62390941-62390941 on Build GRCh38
rs954752649 Chr.20: 62390911-62390911 on Build GRCh38
rs751635728 Chr.20: 62390987-62390987 on Build GRCh38
rs765559284 Chr.20: 62390845-62390845 on Build GRCh38
rs889275037 Chr.20: 62390880-62390880 on Build GRCh38
rs556671748 Chr.20: 62390894-62390894 on Build GRCh38
rs955944816 Chr.20: 62391032-62391032 on Build GRCh38
rs932387746 Chr.20: 62390861-62390861 on Build GRCh38
rs765219271 Chr.20: 62390974-62390974 on Build GRCh38
rs375213613 Chr.20: 62391003-62391003 on Build GRCh38
rs751037345 Chr.20: 62390991-62390991 on Build GRCh38
rs893492445 Chr.20: 62390862-62390862 on Build GRCh38
rs199572235 Chr.20: 62391012-62391012 on Build GRCh38
rs531941270 Chr.20: 62390900-62390900 on Build GRCh38
rs186661638 Chr.20: 62390839-62390839 on Build GRCh38
rs191001143 Chr.20: 62390844-62390844 on Build GRCh38
rs1029456490 Chr.20: 62390903-62390903 on Build GRCh38
rs201904252 Chr.20: 62390928-62390928 on Build GRCh38
rs760108664 Chr.20: 62390897-62390897 on Build GRCh38
rs141862133 Chr.20: 62391046-62391046 on Build GRCh38
rs200133603 Chr.20: 62391010-62391010 on Build GRCh38
rs760347786 Chr.20: 62391041-62391041 on Build GRCh38
rs201067076 Chr.20: 62390998-62390998 on Build GRCh38
rs754984687 Chr.20: 62390988-62390988 on Build GRCh38
rs753618373 Chr.20: 62390942-62390942 on Build GRCh38
rs757199080 Chr.20: 62390954-62390954 on Build GRCh38
rs369535488 Chr.20: 62391018-62391018 on Build GRCh38
rs775062048 Chr.20: 62391013-62391013 on Build GRCh38
rs971597304 Chr.20: 62391001-62391001 on Build GRCh38
rs749662209 Chr.20: 62391008-62391008 on Build GRCh38
rs571998953 Chr.20: 62390927-62390927 on Build GRCh38
rs116729871 Chr.20: 62390840-62390840 on Build GRCh38
rs777409488 Chr.20: 62390899-62390899 on Build GRCh38

View Full Product Details