Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210477637-210477910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
UniGene
Hs.149252
Gene Chromosome Location
Chr.2: 210477682-210679107 on Build GRCh38
Transcripts
NM_001122633.2 XM_011510640.1 XM_011510643.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
719468 c.3+1G>A p.? Chr.2: 210477767-210477767 on Build GRCh38
1631794 c.3+8T>G p.? Chr.2: 210477774-210477774 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs990106910 Chr.2: 210477709-210477710 on Build GRCh38
rs368089210 Chr.2: 210477724-210477724 on Build GRCh38
rs939084908 Chr.2: 210477861-210477861 on Build GRCh38
rs981892575 Chr.2: 210477802-210477802 on Build GRCh38
rs762495642 Chr.2: 210477759-210477759 on Build GRCh38
rs111491997 Chr.2: 210477748-210477748 on Build GRCh38
rs746792298 Chr.2: 210477725-210477725 on Build GRCh38
rs780147302 Chr.2: 210477685-210477685 on Build GRCh38
rs745458954 Chr.2: 210477744-210477744 on Build GRCh38
rs770701357 Chr.2: 210477739-210477739 on Build GRCh38
rs763703546 Chr.2: 210477765-210477765 on Build GRCh38
rs927646465 Chr.2: 210477810-210477810 on Build GRCh38
rs1009029781 Chr.2: 210477686-210477686 on Build GRCh38
rs573133263 Chr.2: 210477690-210477690 on Build GRCh38
rs541490267 Chr.2: 210477869-210477869 on Build GRCh38
rs967487894 Chr.2: 210477694-210477694 on Build GRCh38
rs540925030 Chr.2: 210477667-210477667 on Build GRCh38
rs375555180 Chr.2: 210477713-210477713 on Build GRCh38
rs774927560 Chr.2: 210477756-210477756 on Build GRCh38
rs776620867 Chr.2: 210477743-210477743 on Build GRCh38
rs997045602 Chr.2: 210477670-210477670 on Build GRCh38
rs759322350 Chr.2: 210477747-210477747 on Build GRCh38
rs563233926 Chr.2: 210477770-210477770 on Build GRCh38
rs977107832 Chr.2: 210477700-210477700 on Build GRCh38
rs750805386 Chr.2: 210477814-210477814 on Build GRCh38
rs562721572 Chr.2: 210477771-210477771 on Build GRCh38
rs1032922459 Chr.2: 210477710-210477710 on Build GRCh38
rs187487015 Chr.2: 210477689-210477689 on Build GRCh38
rs1054819481 Chr.2: 210477886-210477886 on Build GRCh38
rs1028476582 Chr.2: 210477672-210477672 on Build GRCh38
rs773855540 Chr.2: 210477795-210477795 on Build GRCh38
rs17552879 Chr.2: 210477778-210477778 on Build GRCh38
rs767714789 Chr.2: 210477780-210477780 on Build GRCh38

View Full Product Details