Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93928388-93928649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM181A
Entrez Gene ID
90050
Gene Name
family with sequence similarity 181 member A
Gene Aliases
C14orf152
UniGene
Hs.525550
Gene Chromosome Location
Chr.14: 93918894-93929608 on Build GRCh38
Transcripts
NM_001207071.1 NM_001207072.1 NM_001207073.1 NM_001207074.1 NM_138344.4 XM_011537297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3356721 c.356G>A p.R119Q Chr.14: 93928455-93928455 on Build GRCh38
4401950 c.520G>A p.E174K Chr.14: 93928619-93928619 on Build GRCh38
7097204 c.355C>T p.R119W Chr.14: 93928454-93928454 on Build GRCh38
292861 c.326G>A p.R109H Chr.14: 93928425-93928425 on Build GRCh38
7203418 c.410G>A p.R137Q Chr.14: 93928509-93928509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370096246 Chr.14: 93928596-93928596 on Build GRCh38
rs752651380 Chr.14: 93928593-93928593 on Build GRCh38
rs777505964 Chr.14: 93928495-93928495 on Build GRCh38
rs139972787 Chr.14: 93928446-93928446 on Build GRCh38
rs1034025317 Chr.14: 93928585-93928585 on Build GRCh38
rs750566102 Chr.14: 93928547-93928547 on Build GRCh38
rs780847444 Chr.14: 93928562-93928562 on Build GRCh38
rs749908917 Chr.14: 93928603-93928603 on Build GRCh38
rs749797057 Chr.14: 93928566-93928566 on Build GRCh38
rs142299390 Chr.14: 93928454-93928454 on Build GRCh38
rs767570265 Chr.14: 93928545-93928545 on Build GRCh38
rs948209001 Chr.14: 93928493-93928494 on Build GRCh38
rs754966355 Chr.14: 93928462-93928462 on Build GRCh38
rs377025207 Chr.14: 93928580-93928580 on Build GRCh38
rs145475417 Chr.14: 93928565-93928565 on Build GRCh38
rs756906691 Chr.14: 93928598-93928598 on Build GRCh38
rs777310102 Chr.14: 93928434-93928434 on Build GRCh38
rs577262012 Chr.14: 93928597-93928597 on Build GRCh38
rs757033068 Chr.14: 93928559-93928559 on Build GRCh38
rs770492169 Chr.14: 93928513-93928513 on Build GRCh38
rs754842393 Chr.14: 93928550-93928550 on Build GRCh38
rs34220325 Chr.14: 93928583-93928583 on Build GRCh38
rs377419146 Chr.14: 93928468-93928468 on Build GRCh38
rs371991985 Chr.14: 93928570-93928570 on Build GRCh38
rs777470544 Chr.14: 93928555-93928555 on Build GRCh38
rs202170145 Chr.14: 93928556-93928556 on Build GRCh38
rs774597064 Chr.14: 93928540-93928540 on Build GRCh38
rs764179723 Chr.14: 93928529-93928529 on Build GRCh38
rs199998912 Chr.14: 93928552-93928552 on Build GRCh38
rs199682227 Chr.14: 93928509-93928509 on Build GRCh38
rs758244029 Chr.14: 93928553-93928553 on Build GRCh38
rs774635941 Chr.14: 93928569-93928569 on Build GRCh38
rs199921751 Chr.14: 93928622-93928622 on Build GRCh38
rs1038294741 Chr.14: 93928470-93928470 on Build GRCh38
rs765046054 Chr.14: 93928592-93928592 on Build GRCh38
rs769039344 Chr.14: 93928567-93928567 on Build GRCh38
rs757932412 Chr.14: 93928487-93928487 on Build GRCh38
rs762848170 Chr.14: 93928594-93928594 on Build GRCh38
rs781008808 Chr.14: 93928602-93928602 on Build GRCh38
rs762070225 Chr.14: 93928543-93928543 on Build GRCh38
rs201033914 Chr.14: 93928499-93928499 on Build GRCh38
rs775654430 Chr.14: 93928515-93928515 on Build GRCh38
rs139773135 Chr.14: 93928472-93928472 on Build GRCh38
rs371120548 Chr.14: 93928508-93928508 on Build GRCh38
rs771516062 Chr.14: 93928424-93928424 on Build GRCh38
rs779476039 Chr.14: 93928605-93928605 on Build GRCh38
rs746462208 Chr.14: 93928498-93928498 on Build GRCh38
rs146784848 Chr.14: 93928455-93928455 on Build GRCh38
rs764387512 Chr.14: 93928453-93928453 on Build GRCh38
rs372111603 Chr.14: 93928445-93928445 on Build GRCh38
rs773217305 Chr.14: 93928591-93928591 on Build GRCh38
rs557191976 Chr.14: 93928459-93928459 on Build GRCh38
rs374170128 Chr.14: 93928519-93928519 on Build GRCh38
rs146610474 Chr.14: 93928629-93928629 on Build GRCh38
rs533878313 Chr.14: 93928563-93928563 on Build GRCh38
rs755614364 Chr.14: 93928604-93928604 on Build GRCh38
rs372090182 Chr.14: 93928452-93928452 on Build GRCh38

View Full Product Details