Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22995977-22996234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf93
Entrez Gene ID
60686
Gene Name
chromosome 14 open reading frame 93
Gene Aliases
-
UniGene
Hs.255874
Gene Chromosome Location
Chr.14: 22986901-23010201 on Build GRCh38
Transcripts
NM_001130706.2 NM_001130708.2 NM_001282968.1 NM_001282969.1 NM_001282970.1 NM_021944.3 XM_005267971.1 XM_005267972.1 XM_006720231.1 XM_006720232.2 XM_011537059.1 XM_011537060.2 XM_011537062.1 XM_017021569.1 XM_017021570.1 XM_017021571.1 XM_017021572.1 XM_017021573.1 XM_017021574.1 XM_017021575.1 XM_017021576.1 XM_017021577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792210 c.650C>T p.P217L Chr.14: 22996216-22996216 on Build GRCh38
6614349 c.638G>A p.R213Q Chr.14: 22996228-22996228 on Build GRCh38
2030780 c.842G>C p.G281A Chr.14: 22996024-22996024 on Build GRCh38
2030781 c.819T>C p.T273T Chr.14: 22996047-22996047 on Build GRCh38
5053864 c.749G>A p.R250H Chr.14: 22996117-22996117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756255543 Chr.14: 22996160-22996160 on Build GRCh38
rs372808152 Chr.14: 22996202-22996202 on Build GRCh38
rs780068233 Chr.14: 22996173-22996173 on Build GRCh38
rs553343257 Chr.14: 22996193-22996193 on Build GRCh38
rs201600502 Chr.14: 22996117-22996117 on Build GRCh38
rs878931408 Chr.14: 22996108-22996108 on Build GRCh38
rs750893238 Chr.14: 22996017-22996017 on Build GRCh38
rs766522355 Chr.14: 22996034-22996034 on Build GRCh38
rs374503197 Chr.14: 22996025-22996025 on Build GRCh38
rs751635736 Chr.14: 22996042-22996042 on Build GRCh38
rs768233107 Chr.14: 22996199-22996199 on Build GRCh38
rs775194846 Chr.14: 22996094-22996094 on Build GRCh38
rs751654219 Chr.14: 22996131-22996131 on Build GRCh38
rs201840699 Chr.14: 22996141-22996141 on Build GRCh38
rs900653587 Chr.14: 22996033-22996033 on Build GRCh38
rs773687054 Chr.14: 22996209-22996209 on Build GRCh38
rs749089379 Chr.14: 22996054-22996054 on Build GRCh38
rs147088064 Chr.14: 22996134-22996134 on Build GRCh38
rs182983042 Chr.14: 22996201-22996201 on Build GRCh38
rs540473017 Chr.14: 22996046-22996046 on Build GRCh38
rs1041914084 Chr.14: 22996028-22996028 on Build GRCh38
rs766785577 Chr.14: 22996127-22996127 on Build GRCh38
rs529811646 Chr.14: 22996169-22996169 on Build GRCh38
rs763586224 Chr.14: 22996125-22996125 on Build GRCh38
rs199624358 Chr.14: 22996016-22996016 on Build GRCh38
rs745634755 Chr.14: 22996168-22996168 on Build GRCh38
rs767762132 Chr.14: 22996135-22996135 on Build GRCh38
rs776382429 Chr.14: 22996106-22996106 on Build GRCh38
rs773183705 Chr.14: 22996092-22996092 on Build GRCh38
rs778831535 Chr.14: 22996167-22996167 on Build GRCh38
rs758809596 Chr.14: 22996019-22996019 on Build GRCh38
rs1009994313 Chr.14: 22996116-22996116 on Build GRCh38
rs369743782 Chr.14: 22996102-22996102 on Build GRCh38
rs755254257 Chr.14: 22996043-22996043 on Build GRCh38
rs745854199 Chr.14: 22996087-22996087 on Build GRCh38
rs953430604 Chr.14: 22996103-22996103 on Build GRCh38
rs757040963 Chr.14: 22996061-22996061 on Build GRCh38
rs773419092 Chr.14: 22996118-22996118 on Build GRCh38
rs150643298 Chr.14: 22996181-22996181 on Build GRCh38
rs762290637 Chr.14: 22996110-22996110 on Build GRCh38
rs141704735 Chr.14: 22996095-22996095 on Build GRCh38
rs763173219 Chr.14: 22996213-22996213 on Build GRCh38
rs574783130 Chr.14: 22996024-22996024 on Build GRCh38
rs762200583 Chr.14: 22996012-22996012 on Build GRCh38
rs377062302 Chr.14: 22996086-22996086 on Build GRCh38

View Full Product Details