Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 1321653-1321900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN6
Entrez Gene ID
27255
Gene Name
contactin 6
Gene Aliases
NB3
UniGene
Hs.387300
Gene Chromosome Location
Chr.3: 978021-1403610 on Build GRCh38
Transcripts
NM_001289080.1 NM_001289081.1 NM_014461.3 XM_005265058.3 XM_011533590.2 XM_011533591.2 XM_017006171.1 XM_017006172.1 XM_017006173.1 XM_017006174.1 XM_017006175.1 XM_017006176.1 XM_017006177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6631839 c.785G>T p.R262I Chr.3: 1321673-1321673 on Build GRCh38
1201688 c.854C>A p.P285Q Chr.3: 1321742-1321742 on Build GRCh38
5660587 c.770C>G p.P257R Chr.3: 1321658-1321658 on Build GRCh38
160092 c.815A>C p.K272T Chr.3: 1321703-1321703 on Build GRCh38
6243072 c.886G>T p.E296* Chr.3: 1321774-1321774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758195551 Chr.3: 1321858-1321858 on Build GRCh38
rs775591980 Chr.3: 1321720-1321720 on Build GRCh38
rs755642212 Chr.3: 1321687-1321687 on Build GRCh38
rs752071699 Chr.3: 1321752-1321752 on Build GRCh38
rs746942934 Chr.3: 1321780-1321780 on Build GRCh38
rs764741013 Chr.3: 1321851-1321851 on Build GRCh38
rs185281214 Chr.3: 1321700-1321700 on Build GRCh38
rs750097513 Chr.3: 1321852-1321852 on Build GRCh38
rs753247872 Chr.3: 1321837-1321837 on Build GRCh38
rs748896656 Chr.3: 1321691-1321691 on Build GRCh38
rs774140967 Chr.3: 1321716-1321722 on Build GRCh38
rs371168661 Chr.3: 1321678-1321678 on Build GRCh38
rs748296604 Chr.3: 1321792-1321792 on Build GRCh38
rs41293401 Chr.3: 1321796-1321796 on Build GRCh38
rs562337434 Chr.3: 1321729-1321729 on Build GRCh38
rs375425277 Chr.3: 1321802-1321802 on Build GRCh38
rs201309309 Chr.3: 1321833-1321833 on Build GRCh38
rs148406106 Chr.3: 1321727-1321727 on Build GRCh38
rs766733914 Chr.3: 1321824-1321824 on Build GRCh38
rs939806418 Chr.3: 1321776-1321776 on Build GRCh38
rs759619716 Chr.3: 1321769-1321769 on Build GRCh38
rs147224770 Chr.3: 1321755-1321755 on Build GRCh38
rs778659800 Chr.3: 1321758-1321758 on Build GRCh38
rs768052985 Chr.3: 1321835-1321835 on Build GRCh38
rs766909657 Chr.3: 1321684-1321684 on Build GRCh38
rs544819651 Chr.3: 1321784-1321784 on Build GRCh38
rs768864425 Chr.3: 1321721-1321721 on Build GRCh38
rs757001842 Chr.3: 1321698-1321698 on Build GRCh38
rs779986260 Chr.3: 1321861-1321861 on Build GRCh38
rs988248049 Chr.3: 1321741-1321741 on Build GRCh38
rs921404133 Chr.3: 1321801-1321801 on Build GRCh38
rs761410258 Chr.3: 1321838-1321838 on Build GRCh38
rs776546906 Chr.3: 1321788-1321788 on Build GRCh38
rs752161155 Chr.3: 1321685-1321685 on Build GRCh38
rs758805504 Chr.3: 1321683-1321683 on Build GRCh38
rs776840689 Chr.3: 1321725-1321725 on Build GRCh38
rs112832949 Chr.3: 1321809-1321809 on Build GRCh38
rs780074087 Chr.3: 1321766-1321766 on Build GRCh38
rs758283823 Chr.3: 1321759-1321759 on Build GRCh38
rs866963872 Chr.3: 1321749-1321749 on Build GRCh38
rs750854152 Chr.3: 1321681-1321681 on Build GRCh38
rs1036885374 Chr.3: 1321795-1321795 on Build GRCh38
rs745717096 Chr.3: 1321702-1321702 on Build GRCh38
rs774763830 Chr.3: 1321828-1321828 on Build GRCh38
rs529488434 Chr.3: 1321754-1321754 on Build GRCh38
rs149244761 Chr.3: 1321703-1321703 on Build GRCh38
rs540929525 Chr.3: 1321708-1321708 on Build GRCh38
rs766820042 Chr.3: 1321745-1321745 on Build GRCh38
rs990536728 Chr.3: 1321711-1321711 on Build GRCh38
rs777348065 Chr.3: 1321689-1321689 on Build GRCh38
rs149799168 Chr.3: 1321816-1321816 on Build GRCh38
rs773414799 Chr.3: 1321743-1321743 on Build GRCh38

View Full Product Details