Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23529551-23529782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EARS2
Entrez Gene ID
124454
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD12, MSE1, gluRS
UniGene
Hs.620541
Gene Chromosome Location
Chr.16: 23522013-23557375 on Build GRCh38
Transcripts
NM_001083614.1 NM_001308211.1 NR_003501.1 XM_011545738.1 XM_011545739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144087 c.1286C>T p.A429V Chr.16: 23529568-23529568 on Build GRCh38
1376796 c.1270C>A p.L424M Chr.16: 23529584-23529584 on Build GRCh38
5628039 c.1223G>A p.G408D Chr.16: 23529631-23529631 on Build GRCh38
6323358 c.1295G>A p.R432Q Chr.16: 23529559-23529559 on Build GRCh38
3279749 c.1206C>T p.I402I Chr.16: 23529759-23529759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752675529 Chr.16: 23529657-23529657 on Build GRCh38
rs977767579 Chr.16: 23529575-23529575 on Build GRCh38
rs779400494 Chr.16: 23529684-23529684 on Build GRCh38
rs921722739 Chr.16: 23529659-23529659 on Build GRCh38
rs781514158 Chr.16: 23529656-23529656 on Build GRCh38
rs750274338 Chr.16: 23529636-23529636 on Build GRCh38
rs759251896 Chr.16: 23529580-23529580 on Build GRCh38
rs961415594 Chr.16: 23529621-23529621 on Build GRCh38
rs199549007 Chr.16: 23529670-23529670 on Build GRCh38
rs771812266 Chr.16: 23529720-23529720 on Build GRCh38
rs753414156 Chr.16: 23529574-23529575 on Build GRCh38
rs771850631 Chr.16: 23529594-23529595 on Build GRCh38
rs200614546 Chr.16: 23529672-23529672 on Build GRCh38
rs746276824 Chr.16: 23529713-23529713 on Build GRCh38
rs763888954 Chr.16: 23529611-23529611 on Build GRCh38
rs549058278 Chr.16: 23529620-23529620 on Build GRCh38
rs768215716 Chr.16: 23529751-23529751 on Build GRCh38
rs919919451 Chr.16: 23529756-23529756 on Build GRCh38
rs774926733 Chr.16: 23529577-23529577 on Build GRCh38
rs923883170 Chr.16: 23529590-23529590 on Build GRCh38
rs368842725 Chr.16: 23529673-23529673 on Build GRCh38
rs773797383 Chr.16: 23529572-23529572 on Build GRCh38
rs371357708 Chr.16: 23529681-23529681 on Build GRCh38
rs745638322 Chr.16: 23529592-23529593 on Build GRCh38
rs760263942 Chr.16: 23529603-23529603 on Build GRCh38
rs749812712 Chr.16: 23529677-23529677 on Build GRCh38
rs756236505 Chr.16: 23529666-23529666 on Build GRCh38
rs749562081 Chr.16: 23529574-23529574 on Build GRCh38
rs1036338305 Chr.16: 23529760-23529760 on Build GRCh38
rs775232277 Chr.16: 23529733-23529733 on Build GRCh38
rs767329476 Chr.16: 23529594-23529594 on Build GRCh38
rs376227171 Chr.16: 23529759-23529759 on Build GRCh38
rs760046153 Chr.16: 23529745-23529745 on Build GRCh38
rs774832573 Chr.16: 23529596-23529596 on Build GRCh38
rs202124923 Chr.16: 23529642-23529642 on Build GRCh38
rs753827689 Chr.16: 23529619-23529619 on Build GRCh38
rs775177860 Chr.16: 23529596-23529597 on Build GRCh38
rs748335490 Chr.16: 23529736-23529738 on Build GRCh38
rs568925129 Chr.16: 23529623-23529623 on Build GRCh38
rs972769786 Chr.16: 23529694-23529694 on Build GRCh38

View Full Product Details