Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81231914-81232169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105371925
Entrez Gene ID
105371925
Gene Name
uncharacterized LOC105371925
Gene Aliases
-
UniGene
Hs.620720
Gene Chromosome Location
Chr.17: 81228707-81232209 on Build GRCh38
Transcripts
NR_135654.1 NM_144679.2 XM_005257066.2 XM_005257067.2 XM_006721709.3 XM_006721712.2 XM_011524355.1 XM_011524356.1 XM_011524357.1 XM_011524358.2 XM_017024202.1 XM_017024203.1 XM_017024204.1

Gene Symbol
TEPSIN
Entrez Gene ID
146705
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Gene Aliases
C17orf56, ENTHD2
UniGene
Hs.631761
Gene Chromosome Location
Chr.17: 81228277-81239092 on Build GRCh38
Transcripts
NR_135654.1 NM_144679.2 XM_005257066.2 XM_005257067.2 XM_006721709.3 XM_006721712.2 XM_011524355.1 XM_011524356.1 XM_011524357.1 XM_011524358.2 XM_017024202.1 XM_017024203.1 XM_017024204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6562625 c.275-4C>G p.? Chr.17: 81232025-81232025 on Build GRCh38
4904059 c.348C>T p.S116S Chr.17: 81231948-81231948 on Build GRCh38
4273139 c.281G>T p.R94M Chr.17: 81232015-81232015 on Build GRCh38
4553122 c.580G>A p.G194S Chr.17: 81231968-81231968 on Build GRCh38
4273137 c.347C>T p.S116F Chr.17: 81231949-81231949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030331367 Chr.17: 81232113-81232113 on Build GRCh38
rs769558985 Chr.17: 81232012-81232012 on Build GRCh38
rs191106052 Chr.17: 81232041-81232041 on Build GRCh38
rs552365485 Chr.17: 81231995-81231995 on Build GRCh38
rs1053561323 Chr.17: 81232128-81232128 on Build GRCh38
rs752604760 Chr.17: 81232056-81232056 on Build GRCh38
rs770569040 Chr.17: 81232025-81232025 on Build GRCh38
rs1012137782 Chr.17: 81231972-81231972 on Build GRCh38
rs780868454 Chr.17: 81231939-81231939 on Build GRCh38
rs777568207 Chr.17: 81232066-81232066 on Build GRCh38
rs151140575 Chr.17: 81231975-81231975 on Build GRCh38
rs762128053 Chr.17: 81231996-81231996 on Build GRCh38
rs761108531 Chr.17: 81231962-81231962 on Build GRCh38
rs770319423 Chr.17: 81231955-81231955 on Build GRCh38
rs532885263 Chr.17: 81232108-81232108 on Build GRCh38
rs773873054 Chr.17: 81231956-81231956 on Build GRCh38
rs200896468 Chr.17: 81231969-81231969 on Build GRCh38
rs997466516 Chr.17: 81232085-81232085 on Build GRCh38
rs150221822 Chr.17: 81232131-81232131 on Build GRCh38
rs563212920 Chr.17: 81231944-81231944 on Build GRCh38
rs777445750 Chr.17: 81232021-81232021 on Build GRCh38
rs764606190 Chr.17: 81231968-81231968 on Build GRCh38
rs747877576 Chr.17: 81231942-81231942 on Build GRCh38
rs201738910 Chr.17: 81231947-81231947 on Build GRCh38
rs766563231 Chr.17: 81232045-81232045 on Build GRCh38
rs767494537 Chr.17: 81232055-81232055 on Build GRCh38
rs762502231 Chr.17: 81231949-81231949 on Build GRCh38
rs749054866 Chr.17: 81232069-81232069 on Build GRCh38
rs747996900 Chr.17: 81232010-81232010 on Build GRCh38
rs772840082 Chr.17: 81231945-81231945 on Build GRCh38
rs751591877 Chr.17: 81232046-81232046 on Build GRCh38
rs954610420 Chr.17: 81232086-81232086 on Build GRCh38
rs34908509 Chr.17: 81231992-81231992 on Build GRCh38
rs528168481 Chr.17: 81232032-81232032 on Build GRCh38
rs368795942 Chr.17: 81232042-81232042 on Build GRCh38
rs758544847 Chr.17: 81231994-81231994 on Build GRCh38
rs532784829 Chr.17: 81231991-81231991 on Build GRCh38
rs765557310 Chr.17: 81232139-81232139 on Build GRCh38
rs754881954 Chr.17: 81232006-81232006 on Build GRCh38
rs748971663 Chr.17: 81232024-81232024 on Build GRCh38
rs759021021 Chr.17: 81232028-81232028 on Build GRCh38
rs371317912 Chr.17: 81231963-81231963 on Build GRCh38
rs746138537 Chr.17: 81231974-81231974 on Build GRCh38
rs199942965 Chr.17: 81232064-81232064 on Build GRCh38
rs746887116 Chr.17: 81231935-81231935 on Build GRCh38
rs551320319 Chr.17: 81232120-81232120 on Build GRCh38
rs755075972 Chr.17: 81232054-81232054 on Build GRCh38
rs758607425 Chr.17: 81232050-81232050 on Build GRCh38
rs773749867 Chr.17: 81232027-81232027 on Build GRCh38
rs11655759 Chr.17: 81231936-81231936 on Build GRCh38
rs879020289 Chr.17: 81231946-81231946 on Build GRCh38
rs987138108 Chr.17: 81232092-81232092 on Build GRCh38
rs781188222 Chr.17: 81232008-81232008 on Build GRCh38
rs77875845 Chr.17: 81231983-81231983 on Build GRCh38
rs762316988 Chr.17: 81232036-81232036 on Build GRCh38
rs943115049 Chr.17: 81232109-81232109 on Build GRCh38
rs113698444 Chr.17: 81232089-81232089 on Build GRCh38

View Full Product Details