Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5216614-5216887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1C4
Entrez Gene ID
1109
Gene Name
aldo-keto reductase family 1, member C4
Gene Aliases
3-alpha-HSD, C11, CDR, CHDR, DD-4, DD4, HAKRA
UniGene
Hs.567245
Gene Chromosome Location
Chr.10: 5196835-5218947 on Build GRCh38
Transcripts
NM_001818.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7135522 c.902G>A p.R301K Chr.10: 5216766-5216766 on Build GRCh38
2146410 c.929T>C p.F310S Chr.10: 5216793-5216793 on Build GRCh38
3438617 c.910C>T p.R304* Chr.10: 5216774-5216774 on Build GRCh38
6129523 c.916G>A p.V306I Chr.10: 5216780-5216780 on Build GRCh38
918827 c.928T>G p.F310V Chr.10: 5216792-5216792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782291916 Chr.10: 5216674-5216674 on Build GRCh38
rs782139163 Chr.10: 5216769-5216769 on Build GRCh38
rs142529153 Chr.10: 5216775-5216775 on Build GRCh38
rs782624554 Chr.10: 5216800-5216800 on Build GRCh38
rs782135898 Chr.10: 5216752-5216752 on Build GRCh38
rs782282343 Chr.10: 5216845-5216845 on Build GRCh38
rs201424763 Chr.10: 5216744-5216744 on Build GRCh38
rs781794131 Chr.10: 5216698-5216698 on Build GRCh38
rs986584843 Chr.10: 5216784-5216784 on Build GRCh38
rs954154722 Chr.10: 5216761-5216761 on Build GRCh38
rs781788408 Chr.10: 5216774-5216774 on Build GRCh38
rs782186398 Chr.10: 5216704-5216704 on Build GRCh38
rs782727831 Chr.10: 5216776-5216776 on Build GRCh38
rs531344596 Chr.10: 5216798-5216798 on Build GRCh38
rs782626293 Chr.10: 5216672-5216672 on Build GRCh38
rs782676546 Chr.10: 5216783-5216783 on Build GRCh38
rs961256248 Chr.10: 5216853-5216853 on Build GRCh38
rs781882270 Chr.10: 5216832-5216833 on Build GRCh38
rs782395452 Chr.10: 5216821-5216821 on Build GRCh38
rs781957017 Chr.10: 5216732-5216732 on Build GRCh38
rs534568556 Chr.10: 5216807-5216807 on Build GRCh38
rs781991900 Chr.10: 5216843-5216843 on Build GRCh38
rs782544054 Chr.10: 5216781-5216781 on Build GRCh38
rs782364741 Chr.10: 5216723-5216723 on Build GRCh38
rs781870024 Chr.10: 5216780-5216780 on Build GRCh38
rs536931267 Chr.10: 5216684-5216684 on Build GRCh38
rs782024904 Chr.10: 5216766-5216766 on Build GRCh38
rs782769364 Chr.10: 5216763-5216763 on Build GRCh38
rs782103594 Chr.10: 5216739-5216739 on Build GRCh38
rs558920470 Chr.10: 5216699-5216699 on Build GRCh38
rs973061093 Chr.10: 5216861-5216861 on Build GRCh38
rs782255336 Chr.10: 5216735-5216735 on Build GRCh38

View Full Product Details