Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61804578-61804851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS1
Entrez Gene ID
3992
Gene Name
fatty acid desaturase 1
Gene Aliases
D5D, FADS6, FADSD5, LLCDL1, TU12
UniGene
Hs.503546
Gene Chromosome Location
Chr.11: 61799625-61817057 on Build GRCh38
Transcripts
NM_013402.4 XM_011545022.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657917 c.868C>T p.R290* Chr.11: 61804699-61804699 on Build GRCh38
5635309 c.854A>G p.Y285C Chr.11: 61804713-61804713 on Build GRCh38
6461935 c.816A>C p.P272P Chr.11: 61804751-61804751 on Build GRCh38
6328690 c.878G>T p.W293L Chr.11: 61804689-61804689 on Build GRCh38
3450801 c.862A>T p.I288F Chr.11: 61804705-61804705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766039999 Chr.11: 61804639-61804639 on Build GRCh38
rs771150406 Chr.11: 61804670-61804670 on Build GRCh38
rs1004018901 Chr.11: 61804781-61804781 on Build GRCh38
rs368642012 Chr.11: 61804613-61804613 on Build GRCh38
rs769194100 Chr.11: 61804792-61804792 on Build GRCh38
rs749758078 Chr.11: 61804786-61804786 on Build GRCh38
rs764151975 Chr.11: 61804830-61804830 on Build GRCh38
rs61695276 Chr.11: 61804801-61804801 on Build GRCh38
rs751651401 Chr.11: 61804832-61804832 on Build GRCh38
rs17856235 Chr.11: 61804753-61804753 on Build GRCh38
rs891011505 Chr.11: 61804769-61804769 on Build GRCh38
rs142276858 Chr.11: 61804699-61804699 on Build GRCh38
rs781597480 Chr.11: 61804754-61804754 on Build GRCh38
rs540146908 Chr.11: 61804815-61804815 on Build GRCh38
rs368350285 Chr.11: 61804799-61804799 on Build GRCh38
rs765159384 Chr.11: 61804684-61804684 on Build GRCh38
rs752575769 Chr.11: 61804816-61804816 on Build GRCh38
rs757506554 Chr.11: 61804751-61804751 on Build GRCh38
rs776157362 Chr.11: 61804810-61804810 on Build GRCh38
rs777968261 Chr.11: 61804725-61804725 on Build GRCh38
rs772942176 Chr.11: 61804783-61804784 on Build GRCh38
rs746177191 Chr.11: 61804773-61804773 on Build GRCh38
rs760291992 Chr.11: 61804637-61804637 on Build GRCh38
rs753625520 Chr.11: 61804674-61804674 on Build GRCh38
rs746224626 Chr.11: 61804809-61804809 on Build GRCh38
rs770500579 Chr.11: 61804805-61804805 on Build GRCh38
rs920713096 Chr.11: 61804623-61804623 on Build GRCh38
rs752563163 Chr.11: 61804687-61804687 on Build GRCh38
rs770072004 Chr.11: 61804776-61804776 on Build GRCh38
rs762959059 Chr.11: 61804817-61804817 on Build GRCh38
rs932297706 Chr.11: 61804610-61804610 on Build GRCh38
rs373761883 Chr.11: 61804634-61804634 on Build GRCh38
rs761577319 Chr.11: 61804785-61804785 on Build GRCh38
rs780529504 Chr.11: 61804780-61804780 on Build GRCh38
rs751739505 Chr.11: 61804736-61804736 on Build GRCh38
rs759250921 Chr.11: 61804682-61804682 on Build GRCh38
rs370431375 Chr.11: 61804648-61804648 on Build GRCh38
rs796197555 Chr.11: 61804648-61804649 on Build GRCh38
rs772028583 Chr.11: 61804735-61804738 on Build GRCh38

View Full Product Details