Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111761100-111761337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX20
Entrez Gene ID
11218
Gene Name
DEAD-box helicase 20
Gene Aliases
DP103, GEMIN3
UniGene
Hs.591405
Gene Chromosome Location
Chr.1: 111755568-111767577 on Build GRCh38
Transcripts
NM_007204.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7123629 c.963-1G>C p.? Chr.1: 111761225-111761225 on Build GRCh38
4970341 c.962+2T>C p.? Chr.1: 111761127-111761127 on Build GRCh38
4563469 c.975G>A p.L325L Chr.1: 111761238-111761238 on Build GRCh38
3984091 c.944T>G p.F315C Chr.1: 111761107-111761107 on Build GRCh38
6503713 c.1018T>C p.S340P Chr.1: 111761281-111761281 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764341690 Chr.1: 111761260-111761260 on Build GRCh38
rs374071540 Chr.1: 111761148-111761148 on Build GRCh38
rs374794942 Chr.1: 111761190-111761190 on Build GRCh38
rs772725238 Chr.1: 111761178-111761178 on Build GRCh38
rs1041982521 Chr.1: 111761238-111761238 on Build GRCh38
rs758652801 Chr.1: 111761156-111761156 on Build GRCh38
rs745780106 Chr.1: 111761208-111761208 on Build GRCh38
rs1057347213 Chr.1: 111761181-111761181 on Build GRCh38
rs748359511 Chr.1: 111761291-111761292 on Build GRCh38
rs770181504 Chr.1: 111761298-111761302 on Build GRCh38
rs568698140 Chr.1: 111761222-111761222 on Build GRCh38
rs763002043 Chr.1: 111761255-111761255 on Build GRCh38
rs747438600 Chr.1: 111761223-111761223 on Build GRCh38
rs145198807 Chr.1: 111761239-111761239 on Build GRCh38
rs756150157 Chr.1: 111761292-111761292 on Build GRCh38
rs762068784 Chr.1: 111761247-111761247 on Build GRCh38
rs17569368 Chr.1: 111761169-111761169 on Build GRCh38
rs372217363 Chr.1: 111761267-111761267 on Build GRCh38
rs752711289 Chr.1: 111761154-111761154 on Build GRCh38
rs944657384 Chr.1: 111761232-111761232 on Build GRCh38
rs1000220277 Chr.1: 111761290-111761290 on Build GRCh38
rs777249440 Chr.1: 111761236-111761236 on Build GRCh38
rs767228360 Chr.1: 111761283-111761283 on Build GRCh38
rs779714618 Chr.1: 111761216-111761216 on Build GRCh38
rs781257765 Chr.1: 111761203-111761203 on Build GRCh38
rs756057137 Chr.1: 111761215-111761215 on Build GRCh38
rs765421485 Chr.1: 111761275-111761275 on Build GRCh38
rs748764511 Chr.1: 111761217-111761217 on Build GRCh38
rs771191854 Chr.1: 111761228-111761228 on Build GRCh38
rs750165086 Chr.1: 111761284-111761284 on Build GRCh38
rs186423567 Chr.1: 111761180-111761180 on Build GRCh38
rs899957346 Chr.1: 111761295-111761295 on Build GRCh38

View Full Product Details