Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 125102833-125103106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AACS
Entrez Gene ID
65985
Gene Name
acetoacetyl-CoA synthetase
Gene Aliases
ACSF1, SUR-5
UniGene
Hs.656073
Gene Chromosome Location
Chr.12: 125065367-125143333 on Build GRCh38
Transcripts
NM_001319839.1 NM_001319840.1 NM_023928.4 XM_011538692.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4505424 c.692C>T p.P231L Chr.12: 125103006-125103006 on Build GRCh38
3687860 c.754A>G p.K252E Chr.12: 125103068-125103068 on Build GRCh38
4835585 c.752C>G p.S251* Chr.12: 125103066-125103066 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569708409 Chr.12: 125102915-125102915 on Build GRCh38
rs78271142 Chr.12: 125102933-125102933 on Build GRCh38
rs1045354756 Chr.12: 125102981-125102981 on Build GRCh38
rs60237232 Chr.12: 125102944-125102944 on Build GRCh38
rs964396791 Chr.12: 125102924-125102924 on Build GRCh38
rs754836022 Chr.12: 125103086-125103086 on Build GRCh38
rs372062541 Chr.12: 125102988-125102988 on Build GRCh38
rs551732352 Chr.12: 125102891-125102891 on Build GRCh38
rs368669260 Chr.12: 125103040-125103040 on Build GRCh38
rs941569701 Chr.12: 125102974-125102974 on Build GRCh38
rs1009972110 Chr.12: 125103066-125103066 on Build GRCh38
rs75448328 Chr.12: 125102931-125102931 on Build GRCh38
rs766258476 Chr.12: 125103058-125103058 on Build GRCh38
rs552840406 Chr.12: 125103015-125103015 on Build GRCh38
rs374355858 Chr.12: 125103047-125103047 on Build GRCh38
rs777020829 Chr.12: 125103005-125103005 on Build GRCh38
rs755304461 Chr.12: 125102955-125102955 on Build GRCh38
rs200531515 Chr.12: 125102998-125102998 on Build GRCh38
rs981548113 Chr.12: 125103006-125103006 on Build GRCh38
rs566929437 Chr.12: 125102993-125102993 on Build GRCh38
rs779221049 Chr.12: 125102961-125102961 on Build GRCh38
rs753641338 Chr.12: 125103081-125103081 on Build GRCh38
rs911191535 Chr.12: 125102907-125102907 on Build GRCh38
rs1012502943 Chr.12: 125102954-125102954 on Build GRCh38
rs79425151 Chr.12: 125102944-125102944 on Build GRCh38
rs536764627 Chr.12: 125102929-125102929 on Build GRCh38
rs771474590 Chr.12: 125102901-125102901 on Build GRCh38
rs534525356 Chr.12: 125103055-125103055 on Build GRCh38
rs878947373 Chr.12: 125102932-125102933 on Build GRCh38
rs748916485 Chr.12: 125103007-125103007 on Build GRCh38
rs376649709 Chr.12: 125103033-125103033 on Build GRCh38
rs771277222 Chr.12: 125103004-125103004 on Build GRCh38
rs767115036 Chr.12: 125103035-125103035 on Build GRCh38
rs748140212 Chr.12: 125102970-125102970 on Build GRCh38
rs978665282 Chr.12: 125102892-125102892 on Build GRCh38
rs150718667 Chr.12: 125103020-125103020 on Build GRCh38
rs752706845 Chr.12: 125102971-125102972 on Build GRCh38
rs755527117 Chr.12: 125102992-125103000 on Build GRCh38
rs41526953 Chr.12: 125103083-125103083 on Build GRCh38
rs760734590 Chr.12: 125103057-125103057 on Build GRCh38

View Full Product Details