Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40462041-40462294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAHD1
Entrez Gene ID
22893
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
UniGene
Hs.22109
Gene Chromosome Location
Chr.15: 40439721-40468242 on Build GRCh38
Transcripts
NM_001301132.1 NM_014952.4 NR_125372.1 XM_011521366.2 XM_011521367.2 XM_011521368.2 XM_011521369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609638 c.1767T>A p.N589K Chr.15: 40462246-40462246 on Build GRCh38
6609637 c.1752T>C p.R584R Chr.15: 40462231-40462231 on Build GRCh38
4054368 c.1735C>T p.R579C Chr.15: 40462214-40462214 on Build GRCh38
4665064 c.1676_1677insA p.R560fs*9 Chr.15: 40462155-40462156 on Build GRCh38
4665063 c.1575C>A p.T525T Chr.15: 40462054-40462054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749648600 Chr.15: 40462080-40462080 on Build GRCh38
rs781186931 Chr.15: 40462146-40462146 on Build GRCh38
rs143007309 Chr.15: 40462236-40462236 on Build GRCh38
rs762888298 Chr.15: 40462231-40462231 on Build GRCh38
rs944527937 Chr.15: 40462177-40462177 on Build GRCh38
rs774266069 Chr.15: 40462233-40462233 on Build GRCh38
rs1038923779 Chr.15: 40462181-40462181 on Build GRCh38
rs778274368 Chr.15: 40462214-40462214 on Build GRCh38
rs765265861 Chr.15: 40462114-40462114 on Build GRCh38
rs762690171 Chr.15: 40462195-40462195 on Build GRCh38
rs1028233291 Chr.15: 40462156-40462156 on Build GRCh38
rs760774172 Chr.15: 40462241-40462241 on Build GRCh38
rs368666699 Chr.15: 40462129-40462129 on Build GRCh38
rs768181332 Chr.15: 40462173-40462173 on Build GRCh38
rs200977266 Chr.15: 40462220-40462220 on Build GRCh38
rs201568757 Chr.15: 40462208-40462208 on Build GRCh38
rs764104056 Chr.15: 40462249-40462249 on Build GRCh38
rs748209295 Chr.15: 40462151-40462151 on Build GRCh38
rs746214980 Chr.15: 40462221-40462221 on Build GRCh38
rs961774486 Chr.15: 40462199-40462199 on Build GRCh38
rs370390459 Chr.15: 40462215-40462215 on Build GRCh38
rs750287065 Chr.15: 40462116-40462116 on Build GRCh38
rs374152126 Chr.15: 40462128-40462128 on Build GRCh38
rs755014028 Chr.15: 40462130-40462130 on Build GRCh38
rs749662166 Chr.15: 40462157-40462157 on Build GRCh38
rs752347404 Chr.15: 40462231-40462232 on Build GRCh38
rs776347545 Chr.15: 40462185-40462185 on Build GRCh38
rs199812510 Chr.15: 40462189-40462189 on Build GRCh38
rs775012276 Chr.15: 40462169-40462169 on Build GRCh38
rs771661919 Chr.15: 40462081-40462081 on Build GRCh38
rs761900037 Chr.15: 40462113-40462113 on Build GRCh38
rs770756651 Chr.15: 40462232-40462232 on Build GRCh38
rs140779269 Chr.15: 40462166-40462166 on Build GRCh38
rs766129464 Chr.15: 40462200-40462200 on Build GRCh38
rs771384339 Chr.15: 40462222-40462223 on Build GRCh38
rs936250730 Chr.15: 40462256-40462256 on Build GRCh38
rs767102133 Chr.15: 40462230-40462231 on Build GRCh38
rs544026902 Chr.15: 40462074-40462074 on Build GRCh38
rs763496269 Chr.15: 40462220-40462222 on Build GRCh38
rs756681057 Chr.15: 40462152-40462152 on Build GRCh38
rs773283717 Chr.15: 40462230-40462230 on Build GRCh38
rs370342543 Chr.15: 40462108-40462108 on Build GRCh38
rs973634298 Chr.15: 40462143-40462143 on Build GRCh38
rs773508937 Chr.15: 40462220-40462234 on Build GRCh38
rs560381001 Chr.15: 40462172-40462172 on Build GRCh38
rs760136146 Chr.15: 40462093-40462093 on Build GRCh38
rs757577200 Chr.15: 40462216-40462216 on Build GRCh38
rs779258465 Chr.15: 40462270-40462270 on Build GRCh38
rs780974137 Chr.15: 40462224-40462224 on Build GRCh38
rs563191486 Chr.15: 40462209-40462209 on Build GRCh38
rs1040703784 Chr.15: 40462073-40462073 on Build GRCh38
rs753850867 Chr.15: 40462254-40462254 on Build GRCh38
rs770279191 Chr.15: 40462219-40462230 on Build GRCh38
rs752866319 Chr.15: 40462239-40462239 on Build GRCh38
rs769391806 Chr.15: 40462229-40462229 on Build GRCh38
rs759073538 Chr.15: 40462226-40462231 on Build GRCh38
rs546204624 Chr.15: 40462190-40462190 on Build GRCh38
rs371637973 Chr.15: 40462154-40462154 on Build GRCh38
rs971210743 Chr.15: 40462232-40462237 on Build GRCh38
rs768466577 Chr.15: 40462223-40462223 on Build GRCh38
rs758192890 Chr.15: 40462127-40462127 on Build GRCh38
rs759220202 Chr.15: 40462235-40462235 on Build GRCh38
rs756162183 Chr.15: 40462212-40462212 on Build GRCh38
rs757849385 Chr.15: 40462259-40462259 on Build GRCh38
rs920569138 Chr.15: 40462144-40462144 on Build GRCh38
rs989396316 Chr.15: 40462264-40462264 on Build GRCh38
rs144296883 Chr.15: 40462109-40462109 on Build GRCh38
rs748457712 Chr.15: 40462214-40462225 on Build GRCh38
rs913375369 Chr.15: 40462269-40462269 on Build GRCh38
rs751740627 Chr.15: 40462202-40462202 on Build GRCh38
rs773873466 Chr.15: 40462225-40462226 on Build GRCh38
rs373800970 Chr.15: 40462226-40462226 on Build GRCh38
rs980263675 Chr.15: 40462234-40462234 on Build GRCh38
rs761380705 Chr.15: 40462186-40462186 on Build GRCh38
rs773624351 Chr.15: 40462075-40462075 on Build GRCh38

View Full Product Details