Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65182673-65182946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN1
Entrez Gene ID
823
Gene Name
calpain 1
Gene Aliases
CANP, CANP1, CANPL1, SPG76, muCANP, muCL
UniGene
Hs.502842
Gene Chromosome Location
Chr.11: 65181215-65212006 on Build GRCh38
Transcripts
NM_001198868.1 NM_001198869.1 NM_005186.3 NR_040008.1 XM_006718698.1 XM_011545292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880458 c.189G>A p.P63P Chr.11: 65182890-65182890 on Build GRCh38
4610910 c.183delC p.P63fs*113 Chr.11: 65182884-65182884 on Build GRCh38
4833390 c.213G>A p.K71K Chr.11: 65182914-65182914 on Build GRCh38
7107022 c.231C>A p.S77S Chr.11: 65182932-65182932 on Build GRCh38
4835470 c.127G>C p.E43Q Chr.11: 65182828-65182828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867375600 Chr.11: 65182706-65182706 on Build GRCh38
rs776728125 Chr.11: 65182871-65182871 on Build GRCh38
rs761450074 Chr.11: 65182886-65182886 on Build GRCh38
rs763840544 Chr.11: 65182725-65182725 on Build GRCh38
rs17881440 Chr.11: 65182905-65182905 on Build GRCh38
rs759639895 Chr.11: 65182875-65182875 on Build GRCh38
rs781338229 Chr.11: 65182881-65182881 on Build GRCh38
rs867213413 Chr.11: 65182812-65182812 on Build GRCh38
rs376855071 Chr.11: 65182721-65182721 on Build GRCh38
rs765399122 Chr.11: 65182884-65182884 on Build GRCh38
rs374461873 Chr.11: 65182837-65182837 on Build GRCh38
rs760603884 Chr.11: 65182696-65182696 on Build GRCh38
rs761299893 Chr.11: 65182866-65182866 on Build GRCh38
rs201318945 Chr.11: 65182922-65182922 on Build GRCh38
rs767415867 Chr.11: 65182887-65182887 on Build GRCh38
rs867076976 Chr.11: 65182789-65182789 on Build GRCh38
rs754042964 Chr.11: 65182722-65182722 on Build GRCh38
rs1006186589 Chr.11: 65182878-65182878 on Build GRCh38
rs768464355 Chr.11: 65182838-65182838 on Build GRCh38
rs770891480 Chr.11: 65182870-65182870 on Build GRCh38
rs912979083 Chr.11: 65182749-65182749 on Build GRCh38
rs564035889 Chr.11: 65182766-65182766 on Build GRCh38
rs750381675 Chr.11: 65182753-65182753 on Build GRCh38
rs773586585 Chr.11: 65182864-65182864 on Build GRCh38
rs866068675 Chr.11: 65182739-65182739 on Build GRCh38
rs780935896 Chr.11: 65182743-65182743 on Build GRCh38
rs986325029 Chr.11: 65182714-65182714 on Build GRCh38
rs759799324 Chr.11: 65182724-65182724 on Build GRCh38
rs201613582 Chr.11: 65182890-65182890 on Build GRCh38
rs17880800 Chr.11: 65182860-65182860 on Build GRCh38
rs758312955 Chr.11: 65182883-65182884 on Build GRCh38
rs778729807 Chr.11: 65182841-65182841 on Build GRCh38
rs748403811 Chr.11: 65182844-65182844 on Build GRCh38
rs752934003 Chr.11: 65182885-65182885 on Build GRCh38
rs754549070 Chr.11: 65182909-65182909 on Build GRCh38
rs751320042 Chr.11: 65182728-65182728 on Build GRCh38
rs1055185439 Chr.11: 65182723-65182723 on Build GRCh38
rs1047886701 Chr.11: 65182790-65182790 on Build GRCh38
rs374112243 Chr.11: 65182695-65182695 on Build GRCh38
rs748927631 Chr.11: 65182834-65182834 on Build GRCh38
rs765955979 Chr.11: 65182901-65182901 on Build GRCh38
rs370263547 Chr.11: 65182803-65182803 on Build GRCh38
rs989863767 Chr.11: 65182729-65182729 on Build GRCh38
rs756128172 Chr.11: 65182894-65182894 on Build GRCh38
rs532996410 Chr.11: 65182772-65182772 on Build GRCh38

View Full Product Details