Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35643314-35643543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV2
Entrez Gene ID
2116
Gene Name
ETS variant 2
Gene Aliases
ER71, ETSRP71
UniGene
Hs.194061
Gene Chromosome Location
Chr.19: 35641175-35644871 on Build GRCh38
Transcripts
NM_001300974.1 NM_001304549.1 NM_014209.3 XM_005258652.2 XM_011526624.2 XM_017026472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4281756 c.353C>A p.P118H Chr.19: 35643391-35643391 on Build GRCh38
3198283 c.564C>G p.P188P Chr.19: 35643518-35643518 on Build GRCh38
3198281 c.437C>A p.P146H Chr.19: 35643391-35643391 on Build GRCh38
7274450 c.401C>A p.P134H Chr.19: 35643439-35643439 on Build GRCh38
4281760 c.480C>G p.P160P Chr.19: 35643518-35643518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770285920 Chr.19: 35643394-35643394 on Build GRCh38
rs776530931 Chr.19: 35643471-35643471 on Build GRCh38
rs555472653 Chr.19: 35643469-35643469 on Build GRCh38
rs180942338 Chr.19: 35643443-35643443 on Build GRCh38
rs944246096 Chr.19: 35643504-35643504 on Build GRCh38
rs1008490600 Chr.19: 35643364-35643364 on Build GRCh38
rs1050161537 Chr.19: 35643473-35643473 on Build GRCh38
rs755996862 Chr.19: 35643386-35643386 on Build GRCh38
rs773895256 Chr.19: 35643409-35643409 on Build GRCh38
rs753561269 Chr.19: 35643338-35643338 on Build GRCh38
rs890020380 Chr.19: 35643356-35643356 on Build GRCh38
rs907675225 Chr.19: 35643420-35643420 on Build GRCh38
rs55634102 Chr.19: 35643419-35643419 on Build GRCh38
rs1021376953 Chr.19: 35643385-35643385 on Build GRCh38
rs777828394 Chr.19: 35643355-35643355 on Build GRCh38
rs940497620 Chr.19: 35643421-35643421 on Build GRCh38
rs751732564 Chr.19: 35643510-35643510 on Build GRCh38
rs369166558 Chr.19: 35643480-35643480 on Build GRCh38
rs527721076 Chr.19: 35643383-35643383 on Build GRCh38
rs758696604 Chr.19: 35643342-35643342 on Build GRCh38
rs751766994 Chr.19: 35643359-35643359 on Build GRCh38
rs749580572 Chr.19: 35643404-35643404 on Build GRCh38
rs1010343699 Chr.19: 35643523-35643523 on Build GRCh38
rs369210600 Chr.19: 35643479-35643479 on Build GRCh38
rs533272713 Chr.19: 35643410-35643410 on Build GRCh38
rs1010396265 Chr.19: 35643388-35643388 on Build GRCh38
rs570810664 Chr.19: 35643405-35643405 on Build GRCh38
rs780706581 Chr.19: 35643372-35643372 on Build GRCh38
rs1053455539 Chr.19: 35643515-35643515 on Build GRCh38
rs760752815 Chr.19: 35643447-35643447 on Build GRCh38
rs766344019 Chr.19: 35643335-35643335 on Build GRCh38
rs369147603 Chr.19: 35643401-35643401 on Build GRCh38
rs555598535 Chr.19: 35643417-35643417 on Build GRCh38
rs562022066 Chr.19: 35643382-35643382 on Build GRCh38
rs1037577316 Chr.19: 35643440-35643440 on Build GRCh38
rs757352567 Chr.19: 35643360-35643360 on Build GRCh38
rs371063485 Chr.19: 35643518-35643518 on Build GRCh38
rs781194637 Chr.19: 35643369-35643369 on Build GRCh38

View Full Product Details