Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44511910-44512183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM20
Entrez Gene ID
125931
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Gene Aliases
UNQ9366
UniGene
Hs.689632
Gene Chromosome Location
Chr.19: 44503838-44530100 on Build GRCh38
Transcripts
NM_001102597.2 NM_001102598.2 NM_001102599.2 NM_001102600.2 XM_011526429.2 XM_011526430.2 XM_011526431.2 XM_011526432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4528807 c.1562G>A p.R521K Chr.19: 44512027-44512027 on Build GRCh38
6397867 c.1564G>T p.V522F Chr.19: 44512025-44512025 on Build GRCh38
3535611 c.1513A>G p.S505G Chr.19: 44512076-44512076 on Build GRCh38
6333208 c.1572+1G>C p.? Chr.19: 44512016-44512016 on Build GRCh38
7448054 c.1543C>G p.Q515E Chr.19: 44512046-44512046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150222142 Chr.19: 44512056-44512056 on Build GRCh38
rs779884221 Chr.19: 44512105-44512105 on Build GRCh38
rs780618317 Chr.19: 44512022-44512022 on Build GRCh38
rs373362306 Chr.19: 44512034-44512034 on Build GRCh38
rs933806580 Chr.19: 44512153-44512153 on Build GRCh38
rs763284714 Chr.19: 44512047-44512047 on Build GRCh38
rs1029066623 Chr.19: 44511946-44511946 on Build GRCh38
rs975194312 Chr.19: 44512115-44512115 on Build GRCh38
rs756085255 Chr.19: 44511971-44511971 on Build GRCh38
rs755408822 Chr.19: 44512125-44512125 on Build GRCh38
rs576892428 Chr.19: 44511990-44511990 on Build GRCh38
rs753815647 Chr.19: 44512064-44512064 on Build GRCh38
rs758968976 Chr.19: 44511992-44511992 on Build GRCh38
rs778238939 Chr.19: 44512095-44512098 on Build GRCh38
rs575882898 Chr.19: 44512100-44512100 on Build GRCh38
rs757977426 Chr.19: 44512017-44512017 on Build GRCh38
rs781770394 Chr.19: 44512033-44512033 on Build GRCh38
rs370270992 Chr.19: 44512129-44512129 on Build GRCh38
rs764864122 Chr.19: 44512081-44512081 on Build GRCh38
rs756801819 Chr.19: 44511984-44511984 on Build GRCh38
rs544840171 Chr.19: 44512103-44512103 on Build GRCh38
rs761654248 Chr.19: 44512070-44512070 on Build GRCh38
rs377370192 Chr.19: 44511966-44511966 on Build GRCh38
rs200024360 Chr.19: 44512044-44512044 on Build GRCh38
rs187368276 Chr.19: 44512057-44512057 on Build GRCh38
rs780040119 Chr.19: 44511994-44511994 on Build GRCh38
rs989997592 Chr.19: 44511969-44511969 on Build GRCh38
rs1034223990 Chr.19: 44512025-44512025 on Build GRCh38
rs749882055 Chr.19: 44512082-44512082 on Build GRCh38
rs753722907 Chr.19: 44511983-44511983 on Build GRCh38
rs771111605 Chr.19: 44512048-44512048 on Build GRCh38
rs750158921 Chr.19: 44511992-44511992 on Build GRCh38
rs552942831 Chr.19: 44512037-44512037 on Build GRCh38
rs763815858 Chr.19: 44512063-44512063 on Build GRCh38
rs931015714 Chr.19: 44512060-44512060 on Build GRCh38
rs764046537 Chr.19: 44511978-44511978 on Build GRCh38
rs564387349 Chr.19: 44512114-44512114 on Build GRCh38
rs8100718 Chr.19: 44512058-44512058 on Build GRCh38
rs781050014 Chr.19: 44512080-44512080 on Build GRCh38
rs878881623 Chr.19: 44511943-44511943 on Build GRCh38
rs747350028 Chr.19: 44512023-44512023 on Build GRCh38

View Full Product Details