Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46657427-46657700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_005253268.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520496.2 XM_011520499.2 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018607.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4972352 c.695+1G>C p.? Chr.11: 46657623-46657623 on Build GRCh38
4663994 c.620T>A p.I207N Chr.11: 46657547-46657547 on Build GRCh38
927250 c.625G>A p.G209R Chr.11: 46657552-46657552 on Build GRCh38
3791516 c.604G>C p.E202Q Chr.11: 46657531-46657531 on Build GRCh38
4663996 c.678G>C p.M226I Chr.11: 46657605-46657605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377546024 Chr.11: 46657600-46657600 on Build GRCh38
rs766114114 Chr.11: 46657491-46657491 on Build GRCh38
rs763403699 Chr.11: 46657633-46657633 on Build GRCh38
rs371095370 Chr.11: 46657643-46657643 on Build GRCh38
rs753647489 Chr.11: 46657562-46657562 on Build GRCh38
rs988757374 Chr.11: 46657567-46657567 on Build GRCh38
rs759057938 Chr.11: 46657502-46657502 on Build GRCh38
rs764494103 Chr.11: 46657631-46657631 on Build GRCh38
rs752152425 Chr.11: 46657658-46657658 on Build GRCh38
rs764007741 Chr.11: 46657671-46657671 on Build GRCh38
rs781161532 Chr.11: 46657596-46657596 on Build GRCh38
rs142832676 Chr.11: 46657569-46657569 on Build GRCh38
rs773910954 Chr.11: 46657616-46657616 on Build GRCh38
rs756042191 Chr.11: 46657509-46657509 on Build GRCh38
rs149706888 Chr.11: 46657539-46657539 on Build GRCh38
rs201283219 Chr.11: 46657599-46657599 on Build GRCh38
rs770212815 Chr.11: 46657608-46657608 on Build GRCh38
rs982084361 Chr.11: 46657603-46657603 on Build GRCh38
rs780215956 Chr.11: 46657580-46657580 on Build GRCh38
rs370111667 Chr.11: 46657499-46657499 on Build GRCh38
rs759136279 Chr.11: 46657645-46657645 on Build GRCh38
rs200669907 Chr.11: 46657593-46657593 on Build GRCh38
rs541823146 Chr.11: 46657477-46657477 on Build GRCh38
rs373092428 Chr.11: 46657669-46657669 on Build GRCh38
rs752635152 Chr.11: 46657507-46657507 on Build GRCh38
rs767017297 Chr.11: 46657653-46657653 on Build GRCh38
rs754190952 Chr.11: 46657640-46657640 on Build GRCh38
rs777478371 Chr.11: 46657538-46657538 on Build GRCh38
rs759627522 Chr.11: 46657593-46657595 on Build GRCh38
rs927372066 Chr.11: 46657606-46657606 on Build GRCh38
rs527396089 Chr.11: 46657450-46657450 on Build GRCh38
rs145070001 Chr.11: 46657579-46657579 on Build GRCh38
rs148242789 Chr.11: 46657584-46657584 on Build GRCh38
rs754012024 Chr.11: 46657497-46657498 on Build GRCh38
rs766988938 Chr.11: 46657504-46657504 on Build GRCh38

View Full Product Details