Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2210635-2210870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRICD5
Entrez Gene ID
283870
Gene Name
BRICHOS domain containing 5
Gene Aliases
C16orf79
UniGene
Hs.647399
Gene Chromosome Location
Chr.16: 2209253-2213037 on Build GRCh38
Transcripts
NM_182563.3 XM_005255263.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948379 c.52-10G>T p.? Chr.16: 2210660-2210660 on Build GRCh38
4257669 c.29G>A p.R10H Chr.16: 2210805-2210805 on Build GRCh38
3279194 c.29G>A p.R10H Chr.16: 2210805-2210805 on Build GRCh38
4400681 c.13A>G p.S5G Chr.16: 2210821-2210821 on Build GRCh38
3948378 c.52-10G>T p.? Chr.16: 2210660-2210660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776662421 Chr.16: 2210676-2210676 on Build GRCh38
rs761853252 Chr.16: 2210677-2210677 on Build GRCh38
rs561323725 Chr.16: 2210665-2210665 on Build GRCh38
rs763670248 Chr.16: 2210656-2210656 on Build GRCh38
rs746769433 Chr.16: 2210674-2210674 on Build GRCh38
rs745441108 Chr.16: 2210659-2210659 on Build GRCh38
rs781640295 Chr.16: 2210655-2210655 on Build GRCh38
rs1034187147 Chr.16: 2210795-2210795 on Build GRCh38
rs774805682 Chr.16: 2210722-2210722 on Build GRCh38
rs373768566 Chr.16: 2210835-2210835 on Build GRCh38
rs375326659 Chr.16: 2210681-2210681 on Build GRCh38
rs781137540 Chr.16: 2210763-2210763 on Build GRCh38
rs781049601 Chr.16: 2210808-2210808 on Build GRCh38
rs774504467 Chr.16: 2210777-2210777 on Build GRCh38
rs774412981 Chr.16: 2210841-2210841 on Build GRCh38
rs755898729 Chr.16: 2210817-2210817 on Build GRCh38
rs530375997 Chr.16: 2210711-2210711 on Build GRCh38
rs760998117 Chr.16: 2210791-2210791 on Build GRCh38
rs761061115 Chr.16: 2210849-2210849 on Build GRCh38
rs763170182 Chr.16: 2210765-2210766 on Build GRCh38
rs367918197 Chr.16: 2210691-2210691 on Build GRCh38
rs759762365 Chr.16: 2210785-2210785 on Build GRCh38
rs200699563 Chr.16: 2210788-2210788 on Build GRCh38
rs778492315 Chr.16: 2210658-2210658 on Build GRCh38
rs764571839 Chr.16: 2210734-2210734 on Build GRCh38
rs768604890 Chr.16: 2210675-2210675 on Build GRCh38
rs151289378 Chr.16: 2210819-2210819 on Build GRCh38
rs764632788 Chr.16: 2210793-2210793 on Build GRCh38
rs764504847 Chr.16: 2210820-2210820 on Build GRCh38
rs547524671 Chr.16: 2210806-2210806 on Build GRCh38
rs768925201 Chr.16: 2210851-2210851 on Build GRCh38
rs186264784 Chr.16: 2210745-2210745 on Build GRCh38
rs866784062 Chr.16: 2210831-2210831 on Build GRCh38
rs949812821 Chr.16: 2210790-2210790 on Build GRCh38
rs773253025 Chr.16: 2210687-2210688 on Build GRCh38
rs372835934 Chr.16: 2210717-2210717 on Build GRCh38
rs201560440 Chr.16: 2210740-2210740 on Build GRCh38
rs368859455 Chr.16: 2210743-2210743 on Build GRCh38
rs775618022 Chr.16: 2210845-2210845 on Build GRCh38
rs767887487 Chr.16: 2210694-2210694 on Build GRCh38
rs754761690 Chr.16: 2210757-2210757 on Build GRCh38
rs1038748148 Chr.16: 2210720-2210720 on Build GRCh38
rs142411784 Chr.16: 2210801-2210801 on Build GRCh38
rs376743515 Chr.16: 2210843-2210843 on Build GRCh38
rs779902576 Chr.16: 2210668-2210668 on Build GRCh38
rs755381079 Chr.16: 2210654-2210654 on Build GRCh38
rs766598102 Chr.16: 2210685-2210685 on Build GRCh38
rs753138553 Chr.16: 2210700-2210700 on Build GRCh38
rs907035934 Chr.16: 2210783-2210783 on Build GRCh38
rs749352890 Chr.16: 2210771-2210771 on Build GRCh38
rs746683461 Chr.16: 2210742-2210742 on Build GRCh38
rs557727646 Chr.16: 2210842-2210842 on Build GRCh38
rs774592132 Chr.16: 2210689-2210689 on Build GRCh38
rs375848463 Chr.16: 2210737-2210737 on Build GRCh38
rs770937860 Chr.16: 2210836-2210836 on Build GRCh38
rs771027960 Chr.16: 2210776-2210776 on Build GRCh38
rs747953077 Chr.16: 2210816-2210816 on Build GRCh38
rs767796924 Chr.16: 2210787-2210787 on Build GRCh38
rs756642039 Chr.16: 2210701-2210701 on Build GRCh38
rs372274139 Chr.16: 2210657-2210657 on Build GRCh38
rs935825995 Chr.16: 2210712-2210712 on Build GRCh38
rs925818305 Chr.16: 2210683-2210683 on Build GRCh38
rs201498388 Chr.16: 2210769-2210769 on Build GRCh38
rs941473417 Chr.16: 2210714-2210714 on Build GRCh38
rs757878009 Chr.16: 2210803-2210803 on Build GRCh38
rs137984071 Chr.16: 2210805-2210805 on Build GRCh38

View Full Product Details