Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9705877-9706144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE9
Entrez Gene ID
151835
Gene Name
copine family member 9
Gene Aliases
-
UniGene
Hs.121335
Gene Chromosome Location
Chr.3: 9703807-9729908 on Build GRCh38
Transcripts
NM_001308388.1 NM_153635.2 XM_011533386.2 XM_011533388.2 XM_011533389.2 XM_017005747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4120675 c.367C>T p.R123* Chr.3: 9706053-9706053 on Build GRCh38
6999645 c.317C>T p.A106V Chr.3: 9706003-9706003 on Build GRCh38
5584902 c.368G>A p.R123Q Chr.3: 9706054-9706054 on Build GRCh38
4674410 c.341T>G p.I114S Chr.3: 9706027-9706027 on Build GRCh38
6999646 c.317C>T p.A106V Chr.3: 9706003-9706003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957349803 Chr.3: 9706008-9706008 on Build GRCh38
rs1000063463 Chr.3: 9705928-9705928 on Build GRCh38
rs772278173 Chr.3: 9706054-9706054 on Build GRCh38
rs768730017 Chr.3: 9705935-9705935 on Build GRCh38
rs370821548 Chr.3: 9705977-9705977 on Build GRCh38
rs761056809 Chr.3: 9706063-9706063 on Build GRCh38
rs376985087 Chr.3: 9706045-9706045 on Build GRCh38
rs201448136 Chr.3: 9706082-9706082 on Build GRCh38
rs200299439 Chr.3: 9706101-9706101 on Build GRCh38
rs201960457 Chr.3: 9706075-9706075 on Build GRCh38
rs766048705 Chr.3: 9706080-9706080 on Build GRCh38
rs536282059 Chr.3: 9706121-9706121 on Build GRCh38
rs756038232 Chr.3: 9706005-9706005 on Build GRCh38
rs550223484 Chr.3: 9706115-9706115 on Build GRCh38
rs772384087 Chr.3: 9705956-9705956 on Build GRCh38
rs769062047 Chr.3: 9706064-9706064 on Build GRCh38
rs780866444 Chr.3: 9706003-9706003 on Build GRCh38
rs749203560 Chr.3: 9706025-9706025 on Build GRCh38
rs777716714 Chr.3: 9706024-9706024 on Build GRCh38
rs771040328 Chr.3: 9706035-9706035 on Build GRCh38
rs775907745 Chr.3: 9705971-9705971 on Build GRCh38
rs371374062 Chr.3: 9706077-9706077 on Build GRCh38
rs775721880 Chr.3: 9706058-9706058 on Build GRCh38
rs1010928457 Chr.3: 9706013-9706013 on Build GRCh38
rs375060861 Chr.3: 9705995-9705995 on Build GRCh38
rs751327697 Chr.3: 9705996-9705996 on Build GRCh38
rs778596001 Chr.3: 9706093-9706093 on Build GRCh38
rs376291148 Chr.3: 9705933-9705933 on Build GRCh38
rs754666010 Chr.3: 9705999-9705999 on Build GRCh38
rs1020442573 Chr.3: 9706053-9706053 on Build GRCh38
rs778957450 Chr.3: 9706039-9706039 on Build GRCh38
rs762499059 Chr.3: 9705989-9705989 on Build GRCh38
rs749928837 Chr.3: 9705984-9705984 on Build GRCh38
rs752473639 Chr.3: 9706004-9706004 on Build GRCh38
rs764594081 Chr.3: 9705982-9705982 on Build GRCh38
rs374925538 Chr.3: 9706070-9706071 on Build GRCh38
rs199944974 Chr.3: 9706068-9706068 on Build GRCh38
rs752458726 Chr.3: 9706088-9706088 on Build GRCh38
rs1023624080 Chr.3: 9706016-9706017 on Build GRCh38
rs752906945 Chr.3: 9706123-9706123 on Build GRCh38
rs759725512 Chr.3: 9705935-9705935 on Build GRCh38
rs773943756 Chr.3: 9706081-9706081 on Build GRCh38

View Full Product Details