Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66727625-66727884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI2
Entrez Gene ID
1783
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Gene Aliases
DNCLI2, LIC2
UniGene
Hs.369068
Gene Chromosome Location
Chr.16: 66720893-66751828 on Build GRCh38
Transcripts
NM_001286157.1 NM_001323955.1 NM_006141.2 NR_136658.1 XM_017023008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609494 c.1169G>C p.G390A Chr.16: 66727780-66727780 on Build GRCh38
6847599 c.1246G>C p.D416H Chr.16: 66727703-66727703 on Build GRCh38
2993715 c.1244C>T p.P415L Chr.16: 66727705-66727705 on Build GRCh38
3754977 c.1236A>G p.V412V Chr.16: 66727713-66727713 on Build GRCh38
4679816 c.1241delA p.K414fs*24 Chr.16: 66727708-66727708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557294372 Chr.16: 66727710-66727710 on Build GRCh38
rs183897123 Chr.16: 66727826-66727826 on Build GRCh38
rs1057503005 Chr.16: 66727859-66727859 on Build GRCh38
rs924860701 Chr.16: 66727842-66727842 on Build GRCh38
rs756130545 Chr.16: 66727739-66727739 on Build GRCh38
rs779678256 Chr.16: 66727651-66727651 on Build GRCh38
rs1033213362 Chr.16: 66727815-66727815 on Build GRCh38
rs373988381 Chr.16: 66727660-66727660 on Build GRCh38
rs772356583 Chr.16: 66727811-66727811 on Build GRCh38
rs17852315 Chr.16: 66727704-66727704 on Build GRCh38
rs775800498 Chr.16: 66727846-66727846 on Build GRCh38
rs751110569 Chr.16: 66727832-66727832 on Build GRCh38
rs778487216 Chr.16: 66727760-66727760 on Build GRCh38
rs768401402 Chr.16: 66727670-66727670 on Build GRCh38
rs750318833 Chr.16: 66727731-66727731 on Build GRCh38
rs113639428 Chr.16: 66727736-66727736 on Build GRCh38
rs145876994 Chr.16: 66727719-66727719 on Build GRCh38
rs559287417 Chr.16: 66727705-66727705 on Build GRCh38
rs754512706 Chr.16: 66727752-66727752 on Build GRCh38
rs146415268 Chr.16: 66727844-66727844 on Build GRCh38
rs773651007 Chr.16: 66727693-66727693 on Build GRCh38
rs774665629 Chr.16: 66727675-66727675 on Build GRCh38
rs747812153 Chr.16: 66727803-66727803 on Build GRCh38
rs10990 Chr.16: 66727713-66727713 on Build GRCh38
rs778178321 Chr.16: 66727821-66727821 on Build GRCh38
rs748470872 Chr.16: 66727678-66727678 on Build GRCh38
rs779406085 Chr.16: 66727746-66727746 on Build GRCh38
rs903459257 Chr.16: 66727750-66727750 on Build GRCh38
rs376699636 Chr.16: 66727840-66727840 on Build GRCh38
rs201115328 Chr.16: 66727715-66727715 on Build GRCh38
rs974512713 Chr.16: 66727732-66727732 on Build GRCh38
rs767437792 Chr.16: 66727728-66727728 on Build GRCh38
rs371546212 Chr.16: 66727720-66727720 on Build GRCh38
rs763329741 Chr.16: 66727855-66727855 on Build GRCh38
rs376571933 Chr.16: 66727839-66727839 on Build GRCh38
rs772447911 Chr.16: 66727684-66727684 on Build GRCh38

View Full Product Details