Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57115377-57115642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE2
Entrez Gene ID
221184
Gene Name
copine 2
Gene Aliases
COPN2, CPN2
UniGene
Hs.339809
Gene Chromosome Location
Chr.16: 57092543-57147966 on Build GRCh38
Transcripts
NM_152727.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362488 c.416C>T p.A139V Chr.16: 57115531-57115531 on Build GRCh38
3510462 c.368C>T p.S123F Chr.16: 57115483-57115483 on Build GRCh38
971795 c.400C>T p.L134L Chr.16: 57115515-57115515 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542921124 Chr.16: 57115578-57115578 on Build GRCh38
rs765650169 Chr.16: 57115454-57115454 on Build GRCh38
rs376741496 Chr.16: 57115549-57115549 on Build GRCh38
rs1050238080 Chr.16: 57115464-57115464 on Build GRCh38
rs879868792 Chr.16: 57115603-57115603 on Build GRCh38
rs768737741 Chr.16: 57115514-57115514 on Build GRCh38
rs767142851 Chr.16: 57115571-57115571 on Build GRCh38
rs1038315430 Chr.16: 57115580-57115580 on Build GRCh38
rs760720084 Chr.16: 57115433-57115433 on Build GRCh38
rs142941150 Chr.16: 57115406-57115406 on Build GRCh38
rs755637015 Chr.16: 57115459-57115459 on Build GRCh38
rs934080701 Chr.16: 57115462-57115462 on Build GRCh38
rs572404586 Chr.16: 57115541-57115541 on Build GRCh38
rs770852542 Chr.16: 57115550-57115550 on Build GRCh38
rs750108302 Chr.16: 57115575-57115575 on Build GRCh38
rs756858718 Chr.16: 57115447-57115447 on Build GRCh38
rs765761440 Chr.16: 57115560-57115560 on Build GRCh38
rs778811580 Chr.16: 57115516-57115516 on Build GRCh38
rs200813420 Chr.16: 57115581-57115581 on Build GRCh38
rs570225877 Chr.16: 57115398-57115398 on Build GRCh38
rs759909007 Chr.16: 57115559-57115559 on Build GRCh38
rs760316086 Chr.16: 57115467-57115467 on Build GRCh38
rs747946748 Chr.16: 57115602-57115602 on Build GRCh38
rs577218567 Chr.16: 57115437-57115437 on Build GRCh38
rs535037978 Chr.16: 57115443-57115443 on Build GRCh38
rs776782991 Chr.16: 57115557-57115557 on Build GRCh38
rs371227636 Chr.16: 57115452-57115452 on Build GRCh38
rs372664504 Chr.16: 57115493-57115493 on Build GRCh38
rs747995967 Chr.16: 57115523-57115523 on Build GRCh38
rs980218142 Chr.16: 57115409-57115409 on Build GRCh38
rs772405976 Chr.16: 57115531-57115531 on Build GRCh38
rs553367188 Chr.16: 57115478-57115478 on Build GRCh38
rs147712310 Chr.16: 57115421-57115421 on Build GRCh38
rs978055830 Chr.16: 57115453-57115453 on Build GRCh38
rs889643209 Chr.16: 57115479-57115479 on Build GRCh38
rs369587924 Chr.16: 57115568-57115568 on Build GRCh38
rs922729478 Chr.16: 57115456-57115456 on Build GRCh38
rs773630857 Chr.16: 57115534-57115534 on Build GRCh38
rs753079229 Chr.16: 57115445-57115445 on Build GRCh38
rs1045426770 Chr.16: 57115591-57115591 on Build GRCh38
rs779046667 Chr.16: 57115596-57115596 on Build GRCh38
rs762725494 Chr.16: 57115511-57115525 on Build GRCh38
rs753420113 Chr.16: 57115584-57115584 on Build GRCh38
rs375561075 Chr.16: 57115594-57115594 on Build GRCh38
rs750688997 Chr.16: 57115505-57115506 on Build GRCh38
rs775796680 Chr.16: 57115566-57115566 on Build GRCh38

View Full Product Details