Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50078236-50078507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC1
Entrez Gene ID
41
Gene Name
acid sensing ion channel subunit 1
Gene Aliases
ACCN2, ASIC, BNaC2
UniGene
Hs.274361
Gene Chromosome Location
Chr.12: 50057637-50083622 on Build GRCh38
Transcripts
NM_001095.3 NM_001256830.1 NM_020039.3 NR_046389.1 XM_011538350.1 XM_011538351.2 XM_011538352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563729 c.885G>A p.M295I Chr.12: 50078468-50078468 on Build GRCh38
431255 c.866C>T p.T289I Chr.12: 50078449-50078449 on Build GRCh38
4614709 c.850delC p.P285fs*85 Chr.12: 50078433-50078433 on Build GRCh38
6072798 c.915C>T p.Y305Y Chr.12: 50078498-50078498 on Build GRCh38
6072799 c.838C>A p.L280I Chr.12: 50078421-50078421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997865059 Chr.12: 50078263-50078263 on Build GRCh38
rs1051048407 Chr.12: 50078281-50078281 on Build GRCh38
rs368875835 Chr.12: 50078406-50078406 on Build GRCh38
rs866451642 Chr.12: 50078386-50078386 on Build GRCh38
rs765512482 Chr.12: 50078378-50078378 on Build GRCh38
rs752923922 Chr.12: 50078388-50078388 on Build GRCh38
rs556272473 Chr.12: 50078283-50078283 on Build GRCh38
rs1029363062 Chr.12: 50078268-50078268 on Build GRCh38
rs901881463 Chr.12: 50078258-50078258 on Build GRCh38
rs555411411 Chr.12: 50078315-50078315 on Build GRCh38
rs569672912 Chr.12: 50078293-50078293 on Build GRCh38
rs535554865 Chr.12: 50078314-50078314 on Build GRCh38
rs147700425 Chr.12: 50078274-50078274 on Build GRCh38
rs962581120 Chr.12: 50078302-50078302 on Build GRCh38
rs748906702 Chr.12: 50078483-50078483 on Build GRCh38
rs144048625 Chr.12: 50078455-50078455 on Build GRCh38
rs886996161 Chr.12: 50078285-50078285 on Build GRCh38
rs971067056 Chr.12: 50078359-50078359 on Build GRCh38
rs867130198 Chr.12: 50078468-50078468 on Build GRCh38
rs898257458 Chr.12: 50078329-50078329 on Build GRCh38
rs981487167 Chr.12: 50078374-50078374 on Build GRCh38
rs1022652055 Chr.12: 50078344-50078344 on Build GRCh38
rs148652119 Chr.12: 50078467-50078467 on Build GRCh38
rs745427778 Chr.12: 50078465-50078465 on Build GRCh38
rs558458817 Chr.12: 50078364-50078364 on Build GRCh38
rs960581152 Chr.12: 50078366-50078366 on Build GRCh38
rs186246026 Chr.12: 50078330-50078330 on Build GRCh38
rs925992809 Chr.12: 50078479-50078479 on Build GRCh38
rs559558875 Chr.12: 50078396-50078396 on Build GRCh38
rs376372385 Chr.12: 50078389-50078389 on Build GRCh38
rs41291983 Chr.12: 50078394-50078394 on Build GRCh38
rs1017086773 Chr.12: 50078290-50078290 on Build GRCh38
rs539867892 Chr.12: 50078275-50078275 on Build GRCh38
rs140797670 Chr.12: 50078327-50078327 on Build GRCh38

View Full Product Details