Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6681784-6682057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3
Entrez Gene ID
718
Gene Name
complement component 3
Gene Aliases
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p
UniGene
Hs.529053
Gene Chromosome Location
Chr.19: 6677835-6720682 on Build GRCh38
Transcripts
NM_000064.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4547618 c.4271G>A p.G1424D Chr.19: 6682020-6682020 on Build GRCh38
6615968 c.4287C>A p.I1429I Chr.19: 6682004-6682004 on Build GRCh38
4288604 c.4322G>A p.R1441K Chr.19: 6681969-6681969 on Build GRCh38
3540985 c.4335C>T p.I1445I Chr.19: 6681956-6681956 on Build GRCh38
4968492 c.4330C>A p.L1444I Chr.19: 6681961-6681961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973056927 Chr.19: 6681836-6681836 on Build GRCh38
rs755251519 Chr.19: 6681950-6681950 on Build GRCh38
rs771805341 Chr.19: 6681899-6681899 on Build GRCh38
rs200878391 Chr.19: 6681977-6681977 on Build GRCh38
rs7951 Chr.19: 6681980-6681980 on Build GRCh38
rs191489530 Chr.19: 6681943-6681943 on Build GRCh38
rs781520118 Chr.19: 6681953-6681953 on Build GRCh38
rs201597793 Chr.19: 6681914-6681914 on Build GRCh38
rs763380367 Chr.19: 6682025-6682025 on Build GRCh38
rs984274936 Chr.19: 6682023-6682023 on Build GRCh38
rs369993135 Chr.19: 6681935-6681935 on Build GRCh38
rs777880161 Chr.19: 6681959-6681959 on Build GRCh38
rs149627133 Chr.19: 6682004-6682004 on Build GRCh38
rs752850583 Chr.19: 6681956-6681956 on Build GRCh38
rs751912518 Chr.19: 6681947-6681947 on Build GRCh38
rs148695501 Chr.19: 6681934-6681934 on Build GRCh38
rs920371959 Chr.19: 6681847-6681847 on Build GRCh38
rs763510721 Chr.19: 6681941-6681941 on Build GRCh38
rs745800783 Chr.19: 6681895-6681895 on Build GRCh38
rs772763964 Chr.19: 6682024-6682024 on Build GRCh38
rs752000215 Chr.19: 6682028-6682028 on Build GRCh38
rs1055468845 Chr.19: 6681915-6681915 on Build GRCh38
rs764831445 Chr.19: 6681922-6681922 on Build GRCh38
rs147116781 Chr.19: 6681972-6681972 on Build GRCh38
rs1043159379 Chr.19: 6681825-6681825 on Build GRCh38
rs768241258 Chr.19: 6681919-6681919 on Build GRCh38
rs775353799 Chr.19: 6681903-6681903 on Build GRCh38
rs113342628 Chr.19: 6681938-6681938 on Build GRCh38
rs761423793 Chr.19: 6681988-6681988 on Build GRCh38
rs756200539 Chr.19: 6681958-6681958 on Build GRCh38
rs377358917 Chr.19: 6682026-6682026 on Build GRCh38
rs772034590 Chr.19: 6681973-6681973 on Build GRCh38
rs143680252 Chr.19: 6681919-6681919 on Build GRCh38
rs776123692 Chr.19: 6681927-6681927 on Build GRCh38
rs1014475053 Chr.19: 6681971-6681971 on Build GRCh38
rs779838912 Chr.19: 6681974-6681974 on Build GRCh38
rs776738013 Chr.19: 6681811-6681811 on Build GRCh38
rs750932330 Chr.19: 6681940-6681940 on Build GRCh38
rs948764879 Chr.19: 6681922-6681922 on Build GRCh38
rs950466120 Chr.19: 6681849-6681849 on Build GRCh38

View Full Product Details