Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34623192-34623453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3C
Entrez Gene ID
138715
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
UniGene
Hs.534549
Gene Chromosome Location
Chr.9: 34621360-34633112 on Build GRCh38
Transcripts
NM_001017363.1 XM_017014283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3413624 c.862A>G p.K288E Chr.9: 34623428-34623428 on Build GRCh38
5463793 c.842T>C p.L281P Chr.9: 34623448-34623448 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761743728 Chr.9: 34623374-34623374 on Build GRCh38
rs942996105 Chr.9: 34623231-34623231 on Build GRCh38
rs910119161 Chr.9: 34623223-34623223 on Build GRCh38
rs767248558 Chr.9: 34623398-34623398 on Build GRCh38
rs896336497 Chr.9: 34623351-34623351 on Build GRCh38
rs756307539 Chr.9: 34623259-34623259 on Build GRCh38
rs1042948063 Chr.9: 34623330-34623330 on Build GRCh38
rs771425589 Chr.9: 34623426-34623426 on Build GRCh38
rs757939936 Chr.9: 34623339-34623339 on Build GRCh38
rs552693900 Chr.9: 34623352-34623352 on Build GRCh38
rs975378434 Chr.9: 34623243-34623243 on Build GRCh38
rs10972173 Chr.9: 34623341-34623341 on Build GRCh38
rs892571129 Chr.9: 34623266-34623266 on Build GRCh38
rs934031428 Chr.9: 34623246-34623246 on Build GRCh38
rs755883257 Chr.9: 34623412-34623412 on Build GRCh38
rs201083706 Chr.9: 34623420-34623420 on Build GRCh38
rs556114150 Chr.9: 34623348-34623348 on Build GRCh38
rs750197167 Chr.9: 34623411-34623411 on Build GRCh38
rs781557631 Chr.9: 34623434-34623434 on Build GRCh38
rs368665257 Chr.9: 34623418-34623418 on Build GRCh38
rs954865601 Chr.9: 34623214-34623214 on Build GRCh38
rs749302265 Chr.9: 34623357-34623357 on Build GRCh38
rs919855847 Chr.9: 34623324-34623324 on Build GRCh38
rs945880609 Chr.9: 34623328-34623328 on Build GRCh38
rs902592156 Chr.9: 34623388-34623388 on Build GRCh38
rs1000872532 Chr.9: 34623409-34623409 on Build GRCh38
rs922615420 Chr.9: 34623244-34623244 on Build GRCh38
rs779825118 Chr.9: 34623417-34623417 on Build GRCh38
rs973676935 Chr.9: 34623269-34623269 on Build GRCh38
rs747472152 Chr.9: 34623425-34623425 on Build GRCh38

View Full Product Details